Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74626132-74626405 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFWD3
Entrez Gene ID
55159
Gene Name
ring finger and WD repeat domain 3
Gene Aliases
RNF201
UniGene
Hs.567525
Gene Chromosome Location
Chr.16: 74621396-74666892 on Build GRCh38
Transcripts
NM_018124.3 XM_005256021.4 XM_005256022.4 XM_006721228.3 XM_011523191.2 XM_017023391.1 XM_017023392.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7328139 c.2164G>A p.A722T Chr.16: 74626360-74626360 on Build GRCh38
5037993 c.2156G>A p.G719E Chr.16: 74626368-74626368 on Build GRCh38
1629987 c.2152A>G p.T718A Chr.16: 74626372-74626372 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775102603 Chr.16: 74626261-74626261 on Build GRCh38
rs748985693 Chr.16: 74626322-74626322 on Build GRCh38
rs767986935 Chr.16: 74626334-74626334 on Build GRCh38
rs913190178 Chr.16: 74626304-74626304 on Build GRCh38
rs189249656 Chr.16: 74626256-74626256 on Build GRCh38
rs946116299 Chr.16: 74626350-74626350 on Build GRCh38
rs751941595 Chr.16: 74626299-74626299 on Build GRCh38
rs573691157 Chr.16: 74626232-74626232 on Build GRCh38
rs78219119 Chr.16: 74626287-74626287 on Build GRCh38
rs768672277 Chr.16: 74626330-74626330 on Build GRCh38
rs936628482 Chr.16: 74626202-74626202 on Build GRCh38
rs1027618385 Chr.16: 74626238-74626238 on Build GRCh38
rs905337447 Chr.16: 74626227-74626227 on Build GRCh38
rs192028101 Chr.16: 74626159-74626159 on Build GRCh38
rs1055080444 Chr.16: 74626174-74626174 on Build GRCh38
rs35195621 Chr.16: 74626308-74626309 on Build GRCh38
rs770870074 Chr.16: 74626382-74626382 on Build GRCh38
rs772096137 Chr.16: 74626362-74626362 on Build GRCh38
rs1046570531 Chr.16: 74626225-74626225 on Build GRCh38
rs778553579 Chr.16: 74626354-74626354 on Build GRCh38
rs751201026 Chr.16: 74626283-74626283 on Build GRCh38
rs775796008 Chr.16: 74626385-74626385 on Build GRCh38
rs537976869 Chr.16: 74626197-74626197 on Build GRCh38
rs544861544 Chr.16: 74626384-74626384 on Build GRCh38
rs781518976 Chr.16: 74626314-74626314 on Build GRCh38
rs760505640 Chr.16: 74626372-74626372 on Build GRCh38
rs184576735 Chr.16: 74626199-74626199 on Build GRCh38
rs757845409 Chr.16: 74626302-74626302 on Build GRCh38
rs1026691312 Chr.16: 74626216-74626216 on Build GRCh38
rs12933037 Chr.16: 74626276-74626276 on Build GRCh38
rs1003920170 Chr.16: 74626274-74626274 on Build GRCh38
rs750831194 Chr.16: 74626315-74626315 on Build GRCh38
rs1000697529 Chr.16: 74626165-74626165 on Build GRCh38
rs756505489 Chr.16: 74626317-74626317 on Build GRCh38
rs149104426 Chr.16: 74626366-74626366 on Build GRCh38
rs574444181 Chr.16: 74626205-74626205 on Build GRCh38
rs77652772 Chr.16: 74626263-74626263 on Build GRCh38
rs1018553447 Chr.16: 74626282-74626282 on Build GRCh38
rs553509447 Chr.16: 74626213-74626213 on Build GRCh38
rs199731609 Chr.16: 74626370-74626370 on Build GRCh38
rs1002473533 Chr.16: 74626233-74626233 on Build GRCh38
rs779635998 Chr.16: 74626320-74626320 on Build GRCh38
rs976792036 Chr.16: 74626289-74626289 on Build GRCh38
rs78201989 Chr.16: 74626207-74626207 on Build GRCh38
rs113552497 Chr.16: 74626332-74626332 on Build GRCh38
rs372258078 Chr.16: 74626187-74626187 on Build GRCh38

View Full Product Details