Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 55588713-55588952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYGO1
Entrez Gene ID
26108
Gene Name
pygopus family PHD finger 1
Gene Aliases
-
UniGene
Hs.256587
Gene Chromosome Location
Chr.15: 55538884-55588852 on Build GRCh38
Transcripts
NM_015617.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
963080 c.34A>T p.K12* Chr.15: 55588819-55588819 on Build GRCh38
1222837 c.7G>A p.A3T Chr.15: 55588846-55588846 on Build GRCh38
6760809 c.45G>A p.S15S Chr.15: 55588808-55588808 on Build GRCh38
1373628 c.44C>T p.S15L Chr.15: 55588809-55588809 on Build GRCh38
2218395 c.17C>T p.S6F Chr.15: 55588836-55588836 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1992236 Chr.15: 55588913-55588913 on Build GRCh38
rs773559111 Chr.15: 55588870-55588870 on Build GRCh38
rs752195295 Chr.15: 55588885-55588885 on Build GRCh38
rs151084609 Chr.15: 55588847-55588847 on Build GRCh38
rs1026444722 Chr.15: 55588745-55588745 on Build GRCh38
rs768745111 Chr.15: 55588843-55588843 on Build GRCh38
rs746730204 Chr.15: 55588774-55588774 on Build GRCh38
rs766740130 Chr.15: 55588876-55588876 on Build GRCh38
rs1036528307 Chr.15: 55588732-55588732 on Build GRCh38
rs145581831 Chr.15: 55588804-55588804 on Build GRCh38
rs939215143 Chr.15: 55588849-55588849 on Build GRCh38
rs756046012 Chr.15: 55588889-55588889 on Build GRCh38
rs988055453 Chr.15: 55588886-55588893 on Build GRCh38
rs776599970 Chr.15: 55588846-55588846 on Build GRCh38
rs535411788 Chr.15: 55588782-55588782 on Build GRCh38
rs758681711 Chr.15: 55588837-55588837 on Build GRCh38
rs777133047 Chr.15: 55588777-55588777 on Build GRCh38
rs756015134 Chr.15: 55588827-55588827 on Build GRCh38
rs752639263 Chr.15: 55588826-55588826 on Build GRCh38
rs768631133 Chr.15: 55588776-55588776 on Build GRCh38
rs749163962 Chr.15: 55588836-55588836 on Build GRCh38
rs367879050 Chr.15: 55588794-55588794 on Build GRCh38
rs950870273 Chr.15: 55588750-55588750 on Build GRCh38
rs763363093 Chr.15: 55588874-55588874 on Build GRCh38
rs138787258 Chr.15: 55588878-55588878 on Build GRCh38
rs557887675 Chr.15: 55588853-55588853 on Build GRCh38
rs753841258 Chr.15: 55588899-55588899 on Build GRCh38
rs202199749 Chr.15: 55588801-55588801 on Build GRCh38
rs146042777 Chr.15: 55588744-55588744 on Build GRCh38
rs773618263 Chr.15: 55588787-55588787 on Build GRCh38
rs1992237 Chr.15: 55588909-55588909 on Build GRCh38
rs766083252 Chr.15: 55588799-55588799 on Build GRCh38
rs369552373 Chr.15: 55588891-55588891 on Build GRCh38
rs200966319 Chr.15: 55588884-55588884 on Build GRCh38
rs765590650 Chr.15: 55588784-55588784 on Build GRCh38
rs772469785 Chr.15: 55588768-55588768 on Build GRCh38
rs747145375 Chr.15: 55588775-55588775 on Build GRCh38
rs922031748 Chr.15: 55588917-55588917 on Build GRCh38
rs1056764699 Chr.15: 55588897-55588897 on Build GRCh38
rs544381422 Chr.15: 55588812-55588812 on Build GRCh38
rs965053300 Chr.15: 55588796-55588796 on Build GRCh38
rs777600024 Chr.15: 55588835-55588835 on Build GRCh38
rs978152127 Chr.15: 55588811-55588811 on Build GRCh38
rs189288408 Chr.15: 55588733-55588733 on Build GRCh38
rs762559220 Chr.15: 55588789-55588789 on Build GRCh38
rs747239416 Chr.15: 55588839-55588839 on Build GRCh38
rs780366882 Chr.15: 55588838-55588838 on Build GRCh38

View Full Product Details