Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59963078-59963339 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNFT1
Entrez Gene ID
51136
Gene Name
ring finger protein, transmembrane 1
Gene Aliases
PTD016
UniGene
Hs.531701
Gene Chromosome Location
Chr.17: 59952362-59964756 on Build GRCh38
Transcripts
NM_016125.3 NR_002924.3

Gene Symbol
TBC1D3P1-DHX40P1
Entrez Gene ID
653645
Gene Name
TBC1D3P1-DHX40P1 readthrough, transcribed pseudogene
Gene Aliases
DHX40P, DHX40P1, TBC1D3P1
UniGene
Hs.448884
Gene Chromosome Location
Chr.17: 59962363-60019052 on Build GRCh38
Transcripts
NM_016125.3 NR_002924.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
294781 c.10A>G p.N4D Chr.17: 59963220-59963220 on Build GRCh38
1303132 c.114G>A p.M38I Chr.17: 59963227-59963227 on Build GRCh38
6249637 c.9C>A p.A3A Chr.17: 59963221-59963221 on Build GRCh38
6249636 c.120C>A p.A40A Chr.17: 59963221-59963221 on Build GRCh38
125903 c.127C>G p.H43D Chr.17: 59963103-59963103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747544806 Chr.17: 59963162-59963162 on Build GRCh38
rs757540470 Chr.17: 59963135-59963135 on Build GRCh38
rs756082554 Chr.17: 59963122-59963122 on Build GRCh38
rs368407775 Chr.17: 59963222-59963222 on Build GRCh38
rs561995422 Chr.17: 59963126-59963126 on Build GRCh38
rs756990650 Chr.17: 59963256-59963256 on Build GRCh38
rs769590656 Chr.17: 59963252-59963257 on Build GRCh38
rs772154437 Chr.17: 59963156-59963156 on Build GRCh38
rs868796726 Chr.17: 59963172-59963172 on Build GRCh38
rs376215881 Chr.17: 59963219-59963219 on Build GRCh38
rs910862562 Chr.17: 59963165-59963165 on Build GRCh38
rs749019062 Chr.17: 59963132-59963132 on Build GRCh38
rs747850231 Chr.17: 59963236-59963239 on Build GRCh38
rs777102208 Chr.17: 59963182-59963182 on Build GRCh38
rs533421595 Chr.17: 59963282-59963282 on Build GRCh38
rs750651147 Chr.17: 59963271-59963271 on Build GRCh38
rs762048699 Chr.17: 59963198-59963198 on Build GRCh38
rs752208915 Chr.17: 59963216-59963216 on Build GRCh38
rs770679705 Chr.17: 59963200-59963200 on Build GRCh38
rs564335135 Chr.17: 59963241-59963241 on Build GRCh38
rs768994695 Chr.17: 59963166-59963166 on Build GRCh38
rs778998844 Chr.17: 59963136-59963136 on Build GRCh38
rs747068326 Chr.17: 59963307-59963307 on Build GRCh38
rs952428753 Chr.17: 59963120-59963120 on Build GRCh38
rs61751975 Chr.17: 59963203-59963203 on Build GRCh38
rs772808400 Chr.17: 59963286-59963287 on Build GRCh38
rs17850123 Chr.17: 59963188-59963188 on Build GRCh38
rs142393490 Chr.17: 59963205-59963205 on Build GRCh38
rs778713054 Chr.17: 59963261-59963261 on Build GRCh38
rs780128550 Chr.17: 59963304-59963304 on Build GRCh38
rs780040800 Chr.17: 59963158-59963158 on Build GRCh38
rs746007285 Chr.17: 59963142-59963142 on Build GRCh38
rs752693194 Chr.17: 59963121-59963121 on Build GRCh38
rs547736586 Chr.17: 59963212-59963212 on Build GRCh38

View Full Product Details