Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40202341-40202566 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLF
Entrez Gene ID
6018
Gene Name
rearranged L-myc fusion
Gene Aliases
ZN-15L, ZNF292L
UniGene
Hs.205627
Gene Chromosome Location
Chr.1: 40161369-40240921 on Build GRCh38
Transcripts
NM_012421.3 XM_017002016.1 XM_017002017.1 XM_017002018.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6564104 c.751C>T p.Q251* Chr.1: 40202555-40202555 on Build GRCh38
1742445 c.743C>T p.S248L Chr.1: 40202547-40202547 on Build GRCh38
323082 c.694A>T p.T232S Chr.1: 40202498-40202498 on Build GRCh38
4721854 c.732T>G p.I244M Chr.1: 40202536-40202536 on Build GRCh38
1963637 c.620T>C p.I207T Chr.1: 40202424-40202424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756447674 Chr.1: 40202426-40202426 on Build GRCh38
rs772374327 Chr.1: 40202483-40202483 on Build GRCh38
rs778720166 Chr.1: 40202391-40202394 on Build GRCh38
rs545236290 Chr.1: 40202525-40202525 on Build GRCh38
rs778168363 Chr.1: 40202387-40202388 on Build GRCh38
rs538064180 Chr.1: 40202458-40202458 on Build GRCh38
rs761707300 Chr.1: 40202510-40202510 on Build GRCh38
rs745979218 Chr.1: 40202480-40202480 on Build GRCh38
rs141908771 Chr.1: 40202388-40202389 on Build GRCh38
rs778273279 Chr.1: 40202427-40202427 on Build GRCh38
rs753861355 Chr.1: 40202376-40202376 on Build GRCh38
rs751214035 Chr.1: 40202536-40202536 on Build GRCh38
rs760483095 Chr.1: 40202490-40202490 on Build GRCh38
rs775871973 Chr.1: 40202487-40202487 on Build GRCh38
rs756492272 Chr.1: 40202542-40202542 on Build GRCh38
rs776618617 Chr.1: 40202498-40202498 on Build GRCh38
rs913064102 Chr.1: 40202430-40202430 on Build GRCh38
rs375770707 Chr.1: 40202370-40202370 on Build GRCh38
rs757073468 Chr.1: 40202408-40202408 on Build GRCh38
rs56855535 Chr.1: 40202389-40202390 on Build GRCh38
rs774340704 Chr.1: 40202459-40202459 on Build GRCh38
rs369418281 Chr.1: 40202513-40202513 on Build GRCh38
rs557902608 Chr.1: 40202515-40202515 on Build GRCh38
rs142014699 Chr.1: 40202535-40202535 on Build GRCh38
rs753512910 Chr.1: 40202384-40202384 on Build GRCh38
rs200904072 Chr.1: 40202389-40202389 on Build GRCh38
rs371978603 Chr.1: 40202524-40202524 on Build GRCh38
rs375128491 Chr.1: 40202497-40202497 on Build GRCh38
rs748462808 Chr.1: 40202419-40202419 on Build GRCh38
rs959932668 Chr.1: 40202369-40202369 on Build GRCh38
rs749822719 Chr.1: 40202439-40202439 on Build GRCh38

View Full Product Details