Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 135119818-135120051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OLFM1
Entrez Gene ID
10439
Gene Name
olfactomedin 1
Gene Aliases
AMY, NOE1, NOELIN1, OlfA
UniGene
Hs.522484
Gene Chromosome Location
Chr.9: 135075243-135121184 on Build GRCh38
Transcripts
NM_001282611.1 NM_001282612.1 NM_014279.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6733209 c.1104C>T p.Y368Y Chr.9: 135119824-135119824 on Build GRCh38
1460960 c.1186G>A p.V396I Chr.9: 135119960-135119960 on Build GRCh38
6227919 c.1261G>A p.D421N Chr.9: 135120035-135120035 on Build GRCh38
5878625 c.1166C>A p.T389N Chr.9: 135119886-135119886 on Build GRCh38
3214529 c.1210A>G p.T404A Chr.9: 135119984-135119984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200726859 Chr.9: 135119933-135119933 on Build GRCh38
rs554344210 Chr.9: 135119909-135119909 on Build GRCh38
rs116667741 Chr.9: 135119969-135119969 on Build GRCh38
rs1039025372 Chr.9: 135119840-135119840 on Build GRCh38
rs376631762 Chr.9: 135119873-135119873 on Build GRCh38
rs746772781 Chr.9: 135119992-135119992 on Build GRCh38
rs772165996 Chr.9: 135119911-135119911 on Build GRCh38
rs775015349 Chr.9: 135119932-135119932 on Build GRCh38
rs757129566 Chr.9: 135120026-135120026 on Build GRCh38
rs201044878 Chr.9: 135119923-135119923 on Build GRCh38
rs770455954 Chr.9: 135119919-135119919 on Build GRCh38
rs762305455 Chr.9: 135119948-135119948 on Build GRCh38
rs764638062 Chr.9: 135119939-135119939 on Build GRCh38
rs767622547 Chr.9: 135119952-135119952 on Build GRCh38
rs777802254 Chr.9: 135119989-135119989 on Build GRCh38
rs932606104 Chr.9: 135119918-135119918 on Build GRCh38
rs573686651 Chr.9: 135119887-135119887 on Build GRCh38
rs777381464 Chr.9: 135119912-135119912 on Build GRCh38
rs1049289 Chr.9: 135119877-135119877 on Build GRCh38
rs775977726 Chr.9: 135119842-135119842 on Build GRCh38
rs760967687 Chr.9: 135119839-135119839 on Build GRCh38
rs1130517 Chr.9: 135119924-135119924 on Build GRCh38
rs956229703 Chr.9: 135120001-135120001 on Build GRCh38
rs759123127 Chr.9: 135119848-135119848 on Build GRCh38
rs780297390 Chr.9: 135119882-135119882 on Build GRCh38
rs764404975 Chr.9: 135119862-135119862 on Build GRCh38
rs201911261 Chr.9: 135119968-135119968 on Build GRCh38
rs756407740 Chr.9: 135119956-135119956 on Build GRCh38
rs766525119 Chr.9: 135119960-135119960 on Build GRCh38
rs756321906 Chr.9: 135119878-135119878 on Build GRCh38
rs530460435 Chr.9: 135119947-135119947 on Build GRCh38
rs751979513 Chr.9: 135119864-135119864 on Build GRCh38
rs757402710 Chr.9: 135119869-135119869 on Build GRCh38
rs373065602 Chr.9: 135119872-135119872 on Build GRCh38
rs3210142 Chr.9: 135119954-135119954 on Build GRCh38
rs544907602 Chr.9: 135119959-135119959 on Build GRCh38
rs753259000 Chr.9: 135119855-135119855 on Build GRCh38
rs540977890 Chr.9: 135119914-135119914 on Build GRCh38
rs763405294 Chr.9: 135119935-135119935 on Build GRCh38
rs750603298 Chr.9: 135119953-135119953 on Build GRCh38
rs61733032 Chr.9: 135119851-135119851 on Build GRCh38
rs866976997 Chr.9: 135119899-135119899 on Build GRCh38
rs3180564 Chr.9: 135119926-135119926 on Build GRCh38
rs971577481 Chr.9: 135119965-135119965 on Build GRCh38

View Full Product Details