Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 84144173-84144440 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKACB
Entrez Gene ID
5567
Gene Name
protein kinase cAMP-activated catalytic subunit beta
Gene Aliases
PKA C-beta, PKACB
UniGene
Hs.487325
Gene Chromosome Location
Chr.1: 84077975-84238498 on Build GRCh38
Transcripts
NM_001300916.1 NM_002731.3 NM_182948.3 NM_207578.2 XM_005271020.1 XM_017001713.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7112615 c.18_19insCCACC p.C9fs*57 Chr.1: 84144379-84144380 on Build GRCh38
85731 c.7G>T p.A3S Chr.1: 84144368-84144368 on Build GRCh38
5734485 c.68A>C p.E23A Chr.1: 84144429-84144429 on Build GRCh38
6587004 c.67G>C p.E23Q Chr.1: 84144428-84144428 on Build GRCh38
5734486 c.69A>C p.E23D Chr.1: 84144430-84144430 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs747486329 Chr.1: 84144385-84144385 on Build GRCh38
rs113718480 Chr.1: 84144277-84144277 on Build GRCh38
rs535443683 Chr.1: 84144238-84144238 on Build GRCh38
rs747789034 Chr.1: 84144343-84144343 on Build GRCh38
rs997355205 Chr.1: 84144275-84144275 on Build GRCh38
rs777242934 Chr.1: 84144393-84144393 on Build GRCh38
rs112013822 Chr.1: 84144337-84144337 on Build GRCh38
rs368743693 Chr.1: 84144387-84144387 on Build GRCh38
rs200815670 Chr.1: 84144374-84144374 on Build GRCh38
rs778856084 Chr.1: 84144335-84144335 on Build GRCh38
rs923525424 Chr.1: 84144368-84144368 on Build GRCh38
rs534766641 Chr.1: 84144326-84144326 on Build GRCh38
rs778642333 Chr.1: 84144397-84144397 on Build GRCh38
rs555443685 Chr.1: 84144298-84144298 on Build GRCh38
rs1028386119 Chr.1: 84144355-84144355 on Build GRCh38
rs566166385 Chr.1: 84144301-84144301 on Build GRCh38
rs1050603869 Chr.1: 84144414-84144414 on Build GRCh38
rs771517024 Chr.1: 84144347-84144347 on Build GRCh38
rs374988248 Chr.1: 84144324-84144324 on Build GRCh38
rs112540650 Chr.1: 84144308-84144308 on Build GRCh38
rs750475419 Chr.1: 84144413-84144413 on Build GRCh38
rs901287940 Chr.1: 84144208-84144208 on Build GRCh38
rs765643686 Chr.1: 84144408-84144408 on Build GRCh38
rs976078812 Chr.1: 84144225-84144226 on Build GRCh38
rs946073117 Chr.1: 84144212-84144212 on Build GRCh38
rs772715076 Chr.1: 84144383-84144383 on Build GRCh38

View Full Product Details