Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47232966-47233227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USP11
Entrez Gene ID
8237
Gene Name
ubiquitin specific peptidase 11
Gene Aliases
UHX1
UniGene
Hs.171501
Gene Chromosome Location
Chr.X: 47232915-47248328 on Build GRCh38
Transcripts
NM_004651.3

SNP Detail

SNP ID Chromosome Location
rs759925085 Chr.X: 47233032-47233032 on Build GRCh38
rs201561406 Chr.X: 47233039-47233039 on Build GRCh38
rs749048718 Chr.X: 47233191-47233191 on Build GRCh38
rs765440597 Chr.X: 47233058-47233058 on Build GRCh38
rs1016903777 Chr.X: 47233037-47233037 on Build GRCh38
rs779991733 Chr.X: 47233066-47233066 on Build GRCh38
rs777419861 Chr.X: 47233132-47233132 on Build GRCh38
rs751547246 Chr.X: 47233096-47233096 on Build GRCh38
rs761237717 Chr.X: 47233043-47233043 on Build GRCh38
rs377322344 Chr.X: 47233055-47233055 on Build GRCh38
rs984035834 Chr.X: 47233085-47233105 on Build GRCh38
rs868143684 Chr.X: 47233042-47233042 on Build GRCh38
rs367543384 Chr.X: 47232987-47232987 on Build GRCh38
rs944203674 Chr.X: 47233103-47233103 on Build GRCh38
rs201098564 Chr.X: 47233134-47233134 on Build GRCh38
rs769346533 Chr.X: 47233130-47233130 on Build GRCh38
rs764305529 Chr.X: 47233049-47233049 on Build GRCh38
rs918867462 Chr.X: 47233076-47233076 on Build GRCh38
rs779901952 Chr.X: 47232985-47232985 on Build GRCh38
rs780637374 Chr.X: 47233115-47233115 on Build GRCh38
rs758150431 Chr.X: 47233065-47233065 on Build GRCh38
rs149757747 Chr.X: 47233024-47233024 on Build GRCh38
rs371419131 Chr.X: 47233012-47233012 on Build GRCh38
rs781722329 Chr.X: 47233079-47233087 on Build GRCh38
rs754879437 Chr.X: 47233105-47233105 on Build GRCh38
rs775961752 Chr.X: 47233151-47233151 on Build GRCh38
rs768106185 Chr.X: 47233004-47233004 on Build GRCh38
rs867773327 Chr.X: 47233162-47233162 on Build GRCh38
rs750188009 Chr.X: 47233060-47233060 on Build GRCh38
rs1003392081 Chr.X: 47233006-47233006 on Build GRCh38
rs772734666 Chr.X: 47233033-47233033 on Build GRCh38
rs774640027 Chr.X: 47233031-47233031 on Build GRCh38
rs776184063 Chr.X: 47233011-47233011 on Build GRCh38
rs985953642 Chr.X: 47233062-47233062 on Build GRCh38
rs747684938 Chr.X: 47233119-47233119 on Build GRCh38
rs746555658 Chr.X: 47232991-47232991 on Build GRCh38
rs200473032 Chr.X: 47233051-47233051 on Build GRCh38
rs868190084 Chr.X: 47233203-47233203 on Build GRCh38
rs757680565 Chr.X: 47233077-47233088 on Build GRCh38
rs772611974 Chr.X: 47233137-47233137 on Build GRCh38

View Full Product Details