Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 118235466-118235707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35F1
Entrez Gene ID
222553
Gene Name
solute carrier family 35 member F1
Gene Aliases
C6orf169, dJ230I3.1
UniGene
Hs.654841
Gene Chromosome Location
Chr.6: 117907226-118317676 on Build GRCh38
Transcripts
NM_001029858.3 XM_005266865.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
361003 c.439G>T p.V147F Chr.6: 118235598-118235598 on Build GRCh38
6172022 c.386G>T p.W129L Chr.6: 118235545-118235545 on Build GRCh38
6547600 c.421G>T p.E141* Chr.6: 118235580-118235580 on Build GRCh38
3857853 c.382A>C p.R128R Chr.6: 118235541-118235541 on Build GRCh38
3857852 c.379C>T p.R127* Chr.6: 118235538-118235538 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773761917 Chr.6: 118235504-118235504 on Build GRCh38
rs374955983 Chr.6: 118235662-118235662 on Build GRCh38
rs773864511 Chr.6: 118235536-118235536 on Build GRCh38
rs367694719 Chr.6: 118235668-118235668 on Build GRCh38
rs1030022120 Chr.6: 118235572-118235572 on Build GRCh38
rs761498734 Chr.6: 118235657-118235657 on Build GRCh38
rs771528739 Chr.6: 118235560-118235560 on Build GRCh38
rs749588500 Chr.6: 118235505-118235505 on Build GRCh38
rs866598352 Chr.6: 118235567-118235567 on Build GRCh38
rs763987457 Chr.6: 118235628-118235628 on Build GRCh38
rs770687776 Chr.6: 118235519-118235519 on Build GRCh38
rs753732894 Chr.6: 118235643-118235643 on Build GRCh38
rs760651651 Chr.6: 118235566-118235566 on Build GRCh38
rs114837400 Chr.6: 118235671-118235671 on Build GRCh38
rs775424691 Chr.6: 118235562-118235562 on Build GRCh38
rs759102731 Chr.6: 118235538-118235538 on Build GRCh38
rs200553180 Chr.6: 118235493-118235493 on Build GRCh38
rs201078068 Chr.6: 118235522-118235522 on Build GRCh38
rs766636803 Chr.6: 118235665-118235665 on Build GRCh38
rs745950537 Chr.6: 118235539-118235539 on Build GRCh38
rs1006506689 Chr.6: 118235503-118235503 on Build GRCh38
rs866740424 Chr.6: 118235605-118235605 on Build GRCh38

View Full Product Details