Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 74243127-74243356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC2
Entrez Gene ID
5982
Gene Name
replication factor C subunit 2
Gene Aliases
RFC40
UniGene
Hs.647062
Gene Chromosome Location
Chr.7: 74231502-74254458 on Build GRCh38
Transcripts
NM_001278791.1 NM_001278792.1 NM_001278793.1 NM_002914.4 NM_181471.2 XM_006716080.2 XM_017012492.1 XM_017012493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202834 c.523G>C p.D175H Chr.7: 74243158-74243158 on Build GRCh38
5421296 c.450C>G p.A150A Chr.7: 74243231-74243231 on Build GRCh38
4429315 c.441C>T p.T147T Chr.7: 74243240-74243240 on Build GRCh38
4548418 c.446G>A p.G149E Chr.7: 74243235-74243235 on Build GRCh38
5451915 c.481T>C p.Y161H Chr.7: 74243200-74243200 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782704669 Chr.7: 74243242-74243242 on Build GRCh38
rs143077432 Chr.7: 74243201-74243201 on Build GRCh38
rs1042801182 Chr.7: 74243295-74243295 on Build GRCh38
rs782411584 Chr.7: 74243166-74243166 on Build GRCh38
rs782479769 Chr.7: 74243154-74243154 on Build GRCh38
rs782678856 Chr.7: 74243297-74243297 on Build GRCh38
rs200629497 Chr.7: 74243245-74243245 on Build GRCh38
rs782066892 Chr.7: 74243186-74243186 on Build GRCh38
rs377423144 Chr.7: 74243264-74243264 on Build GRCh38
rs782587574 Chr.7: 74243275-74243275 on Build GRCh38
rs915177375 Chr.7: 74243253-74243253 on Build GRCh38
rs373087071 Chr.7: 74243240-74243240 on Build GRCh38
rs782220982 Chr.7: 74243277-74243277 on Build GRCh38
rs782118846 Chr.7: 74243184-74243184 on Build GRCh38
rs926446177 Chr.7: 74243232-74243232 on Build GRCh38
rs782537107 Chr.7: 74243247-74243247 on Build GRCh38
rs200469992 Chr.7: 74243158-74243158 on Build GRCh38
rs781847598 Chr.7: 74243261-74243261 on Build GRCh38
rs782680458 Chr.7: 74243260-74243260 on Build GRCh38
rs139127548 Chr.7: 74243159-74243159 on Build GRCh38
rs989354651 Chr.7: 74243248-74243248 on Build GRCh38
rs959223183 Chr.7: 74243233-74243233 on Build GRCh38
rs146804166 Chr.7: 74243296-74243296 on Build GRCh38
rs782042572 Chr.7: 74243180-74243180 on Build GRCh38
rs945357102 Chr.7: 74243287-74243287 on Build GRCh38
rs781826115 Chr.7: 74243237-74243237 on Build GRCh38

View Full Product Details