Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 82357187-82357438 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC59
Entrez Gene ID
29080
Gene Name
coiled-coil domain containing 59
Gene Aliases
BR22, HSPC128, TAP26
UniGene
Hs.582627
Gene Chromosome Location
Chr.12: 82352304-82358805 on Build GRCh38
Transcripts
NM_014167.4 NR_033192.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5747428 c.155-2A>G p.? Chr.12: 82357271-82357271 on Build GRCh38
4534103 c.204G>A p.K68K Chr.12: 82357220-82357220 on Build GRCh38
3466014 c.173G>A p.R58Q Chr.12: 82357251-82357251 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1007782361 Chr.12: 82357406-82357406 on Build GRCh38
rs934422179 Chr.12: 82357375-82357375 on Build GRCh38
rs909219241 Chr.12: 82357302-82357302 on Build GRCh38
rs769263988 Chr.12: 82357282-82357283 on Build GRCh38
rs149629092 Chr.12: 82357287-82357287 on Build GRCh38
rs12818192 Chr.12: 82357320-82357320 on Build GRCh38
rs769748085 Chr.12: 82357305-82357305 on Build GRCh38
rs777457552 Chr.12: 82357242-82357242 on Build GRCh38
rs398055770 Chr.12: 82357287-82357289 on Build GRCh38
rs775676423 Chr.12: 82357228-82357228 on Build GRCh38
rs2401035 Chr.12: 82357388-82357388 on Build GRCh38
rs199701665 Chr.12: 82357285-82357285 on Build GRCh38
rs138370934 Chr.12: 82357308-82357308 on Build GRCh38
rs915716553 Chr.12: 82357343-82357343 on Build GRCh38
rs949907900 Chr.12: 82357352-82357352 on Build GRCh38
rs1045562461 Chr.12: 82357355-82357355 on Build GRCh38
rs907195672 Chr.12: 82357353-82357356 on Build GRCh38
rs984781426 Chr.12: 82357288-82357288 on Build GRCh38
rs759432434 Chr.12: 82357209-82357209 on Build GRCh38
rs765655707 Chr.12: 82357314-82357316 on Build GRCh38
rs772493909 Chr.12: 82357274-82357274 on Build GRCh38
rs892813654 Chr.12: 82357403-82357403 on Build GRCh38
rs752684922 Chr.12: 82357232-82357232 on Build GRCh38
rs936505463 Chr.12: 82357279-82357279 on Build GRCh38
rs3077481 Chr.12: 82357284-82357286 on Build GRCh38
rs777086687 Chr.12: 82357295-82357295 on Build GRCh38
rs12816738 Chr.12: 82357328-82357328 on Build GRCh38
rs762271916 Chr.12: 82357302-82357303 on Build GRCh38
rs745402383 Chr.12: 82357379-82357379 on Build GRCh38
rs950816865 Chr.12: 82357278-82357278 on Build GRCh38
rs767444064 Chr.12: 82357313-82357313 on Build GRCh38
rs778690208 Chr.12: 82357252-82357252 on Build GRCh38
rs767415058 Chr.12: 82357210-82357210 on Build GRCh38
rs745506195 Chr.12: 82357273-82357273 on Build GRCh38
rs966625252 Chr.12: 82357236-82357236 on Build GRCh38
rs752501352 Chr.12: 82357315-82357315 on Build GRCh38
rs772256609 Chr.12: 82357223-82357224 on Build GRCh38
rs747534553 Chr.12: 82357281-82357281 on Build GRCh38
rs549311411 Chr.12: 82357351-82357351 on Build GRCh38
rs536951247 Chr.12: 82357362-82357362 on Build GRCh38
rs200349482 Chr.12: 82357286-82357286 on Build GRCh38
rs563563497 Chr.12: 82357311-82357311 on Build GRCh38
rs1034421135 Chr.12: 82357365-82357365 on Build GRCh38
rs1017749409 Chr.12: 82357224-82357224 on Build GRCh38
rs747050998 Chr.12: 82357241-82357242 on Build GRCh38
rs756700518 Chr.12: 82357249-82357249 on Build GRCh38
rs10506870 Chr.12: 82357321-82357321 on Build GRCh38
rs762001352 Chr.12: 82357298-82357298 on Build GRCh38
rs921119943 Chr.12: 82357248-82357248 on Build GRCh38
rs756069326 Chr.12: 82357234-82357234 on Build GRCh38
rs769108137 Chr.12: 82357294-82357294 on Build GRCh38
rs1039682282 Chr.12: 82357409-82357409 on Build GRCh38
rs753440706 Chr.12: 82357243-82357243 on Build GRCh38
rs780335986 Chr.12: 82357277-82357277 on Build GRCh38

View Full Product Details