Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 5291146-5291391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZBTB14
Entrez Gene ID
7541
Gene Name
zinc finger and BTB domain containing 14
Gene Aliases
ZF5, ZFP-161, ZFP-5, ZFP161, ZNF478
UniGene
Hs.592340
Gene Chromosome Location
Chr.18: 5289019-5297053 on Build GRCh38
Transcripts
NM_001143823.2 NM_001243702.1 NM_001243704.1 NM_003409.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3378524 c.1029C>T p.I343I Chr.18: 5291179-5291179 on Build GRCh38
3526547 c.1022C>T p.S341L Chr.18: 5291186-5291186 on Build GRCh38
4934485 c.882G>T p.K294N Chr.18: 5291326-5291326 on Build GRCh38
5056075 c.910C>T p.P304S Chr.18: 5291298-5291298 on Build GRCh38
5510944 c.820C>T p.R274W Chr.18: 5291388-5291388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779305024 Chr.18: 5291254-5291254 on Build GRCh38
rs749296377 Chr.18: 5291309-5291309 on Build GRCh38
rs1009768456 Chr.18: 5291271-5291271 on Build GRCh38
rs766905979 Chr.18: 5291199-5291199 on Build GRCh38
rs907906573 Chr.18: 5291206-5291206 on Build GRCh38
rs780154344 Chr.18: 5291306-5291306 on Build GRCh38
rs780684444 Chr.18: 5291295-5291295 on Build GRCh38
rs992822668 Chr.18: 5291200-5291200 on Build GRCh38
rs144708635 Chr.18: 5291354-5291354 on Build GRCh38
rs749766580 Chr.18: 5291311-5291311 on Build GRCh38
rs760626422 Chr.18: 5291188-5291188 on Build GRCh38
rs960148609 Chr.18: 5291193-5291193 on Build GRCh38
rs34402690 Chr.18: 5291305-5291305 on Build GRCh38
rs756971572 Chr.18: 5291279-5291279 on Build GRCh38
rs143791680 Chr.18: 5291302-5291302 on Build GRCh38
rs767606188 Chr.18: 5291178-5291178 on Build GRCh38
rs761492873 Chr.18: 5291335-5291335 on Build GRCh38
rs374218314 Chr.18: 5291339-5291339 on Build GRCh38
rs138277959 Chr.18: 5291224-5291224 on Build GRCh38
rs755346237 Chr.18: 5291246-5291246 on Build GRCh38
rs149008980 Chr.18: 5291269-5291269 on Build GRCh38
rs773269856 Chr.18: 5291341-5291341 on Build GRCh38
rs760818543 Chr.18: 5291353-5291353 on Build GRCh38
rs750568439 Chr.18: 5291181-5291181 on Build GRCh38
rs779444390 Chr.18: 5291365-5291365 on Build GRCh38
rs140694454 Chr.18: 5291323-5291323 on Build GRCh38
rs369177970 Chr.18: 5291334-5291334 on Build GRCh38
rs753898579 Chr.18: 5291371-5291371 on Build GRCh38

View Full Product Details