Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 30366114-30366369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC157
Entrez Gene ID
550631
Gene Name
coiled-coil domain containing 157
Gene Aliases
-
UniGene
Hs.505597
Gene Chromosome Location
Chr.22: 30356635-30378655 on Build GRCh38
Transcripts
NM_001017437.3 NM_001318334.1 NM_001318335.1 XM_006724278.3 XM_006724279.3 XM_006724280.3 XM_011530263.2 XM_011530266.2 XM_011530267.2 XM_011530268.2 XM_011530270.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3764923 c.54G>A p.T18T Chr.22: 30366207-30366207 on Build GRCh38
333389 c.134G>A p.R45H Chr.22: 30366134-30366134 on Build GRCh38
725956 c.62C>T p.S21F Chr.22: 30366215-30366215 on Build GRCh38
6161981 c.180C>T p.D60D Chr.22: 30366180-30366180 on Build GRCh38
6008616 c.140C>G p.A47G Chr.22: 30366140-30366140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752731939 Chr.22: 30366149-30366149 on Build GRCh38
rs758422993 Chr.22: 30366209-30366209 on Build GRCh38
rs41281643 Chr.22: 30366189-30366189 on Build GRCh38
rs558203267 Chr.22: 30366207-30366207 on Build GRCh38
rs766620698 Chr.22: 30366199-30366199 on Build GRCh38
rs775428315 Chr.22: 30366267-30366267 on Build GRCh38
rs766962692 Chr.22: 30366290-30366290 on Build GRCh38
rs779274704 Chr.22: 30366225-30366225 on Build GRCh38
rs375975371 Chr.22: 30366249-30366249 on Build GRCh38
rs377414522 Chr.22: 30366256-30366256 on Build GRCh38
rs1002974402 Chr.22: 30366324-30366324 on Build GRCh38
rs989186826 Chr.22: 30366281-30366281 on Build GRCh38
rs199951971 Chr.22: 30366139-30366139 on Build GRCh38
rs777350775 Chr.22: 30366213-30366213 on Build GRCh38
rs188579166 Chr.22: 30366162-30366162 on Build GRCh38
rs756772183 Chr.22: 30366221-30366221 on Build GRCh38
rs752632130 Chr.22: 30366208-30366208 on Build GRCh38
rs762905873 Chr.22: 30366271-30366271 on Build GRCh38
rs377629910 Chr.22: 30366231-30366231 on Build GRCh38
rs573801267 Chr.22: 30366332-30366332 on Build GRCh38
rs773777523 Chr.22: 30366243-30366243 on Build GRCh38
rs568353300 Chr.22: 30366175-30366175 on Build GRCh38
rs201763455 Chr.22: 30366134-30366134 on Build GRCh38
rs113058207 Chr.22: 30366198-30366198 on Build GRCh38
rs754066462 Chr.22: 30366205-30366205 on Build GRCh38
rs199911862 Chr.22: 30366136-30366136 on Build GRCh38
rs750607091 Chr.22: 30366133-30366133 on Build GRCh38
rs539513309 Chr.22: 30366206-30366206 on Build GRCh38
rs1015097917 Chr.22: 30366317-30366317 on Build GRCh38
rs987402862 Chr.22: 30366333-30366333 on Build GRCh38
rs151177194 Chr.22: 30366191-30366191 on Build GRCh38
rs921789110 Chr.22: 30366312-30366312 on Build GRCh38
rs145073380 Chr.22: 30366222-30366222 on Build GRCh38
rs141865302 Chr.22: 30366244-30366244 on Build GRCh38
rs147104395 Chr.22: 30366245-30366245 on Build GRCh38
rs778897051 Chr.22: 30366145-30366145 on Build GRCh38
rs740223 Chr.22: 30366151-30366151 on Build GRCh38
rs772574462 Chr.22: 30366238-30366238 on Build GRCh38
rs529483498 Chr.22: 30366188-30366188 on Build GRCh38
rs751497200 Chr.22: 30366279-30366279 on Build GRCh38
rs41281641 Chr.22: 30366150-30366150 on Build GRCh38
rs749647847 Chr.22: 30366179-30366179 on Build GRCh38
rs528762178 Chr.22: 30366157-30366157 on Build GRCh38
rs147562085 Chr.22: 30366223-30366223 on Build GRCh38
rs775039007 Chr.22: 30366279-30366280 on Build GRCh38
rs948351480 Chr.22: 30366341-30366341 on Build GRCh38
rs555691517 Chr.22: 30366300-30366300 on Build GRCh38
rs751107362 Chr.22: 30366218-30366218 on Build GRCh38
rs202240153 Chr.22: 30366291-30366291 on Build GRCh38
rs764107166 Chr.22: 30366273-30366273 on Build GRCh38
rs897873540 Chr.22: 30366318-30366318 on Build GRCh38
rs756756781 Chr.22: 30366288-30366288 on Build GRCh38
rs370242340 Chr.22: 30366292-30366292 on Build GRCh38

View Full Product Details