Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50374866-50375139 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
UniGene
Hs.38132
Gene Chromosome Location
Chr.X: 50365407-50470850 on Build GRCh38
Transcripts
NM_001013742.3 XM_017029268.1

SNP Detail

SNP ID Chromosome Location
rs782219540 Chr.X: 50374904-50374904 on Build GRCh38
rs781787603 Chr.X: 50374957-50374957 on Build GRCh38
rs376854796 Chr.X: 50375003-50375003 on Build GRCh38
rs7473051 Chr.X: 50374899-50374899 on Build GRCh38
rs782030192 Chr.X: 50375117-50375117 on Build GRCh38
rs782256342 Chr.X: 50374974-50374974 on Build GRCh38
rs372217757 Chr.X: 50374990-50374990 on Build GRCh38
rs184591941 Chr.X: 50375094-50375094 on Build GRCh38
rs782592163 Chr.X: 50374961-50374961 on Build GRCh38
rs192043707 Chr.X: 50375037-50375037 on Build GRCh38
rs782554813 Chr.X: 50374947-50374947 on Build GRCh38
rs781784401 Chr.X: 50374996-50374996 on Build GRCh38
rs781955690 Chr.X: 50374956-50374957 on Build GRCh38
rs782285579 Chr.X: 50375004-50375004 on Build GRCh38
rs782495355 Chr.X: 50374958-50374958 on Build GRCh38
rs782003748 Chr.X: 50375071-50375071 on Build GRCh38
rs782591580 Chr.X: 50374948-50374948 on Build GRCh38
rs371705438 Chr.X: 50375036-50375036 on Build GRCh38
rs370174837 Chr.X: 50375073-50375073 on Build GRCh38
rs781862419 Chr.X: 50374940-50374940 on Build GRCh38
rs782814351 Chr.X: 50375082-50375082 on Build GRCh38
rs782709247 Chr.X: 50374993-50374993 on Build GRCh38
rs1029613548 Chr.X: 50374909-50374909 on Build GRCh38

View Full Product Details