Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 4696999-4697244 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC37L1
Entrez Gene ID
55664
Gene Name
cell division cycle 37 like 1
Gene Aliases
CDC37B, HARC
UniGene
Hs.666439
Gene Chromosome Location
Chr.9: 4679553-4708399 on Build GRCh38
Transcripts
NM_017913.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3907502 c.529G>C p.D177H Chr.9: 4697116-4697116 on Build GRCh38
455993 c.607C>T p.H203Y Chr.9: 4697194-4697194 on Build GRCh38
5983794 c.561C>A p.Y187* Chr.9: 4697148-4697148 on Build GRCh38
4791606 c.514T>G p.L172V Chr.9: 4697101-4697101 on Build GRCh38
1109330 c.539G>T p.R180I Chr.9: 4697126-4697126 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766876467 Chr.9: 4697133-4697133 on Build GRCh38
rs775996289 Chr.9: 4697103-4697103 on Build GRCh38
rs1005594778 Chr.9: 4697083-4697083 on Build GRCh38
rs750004119 Chr.9: 4697118-4697118 on Build GRCh38
rs200881971 Chr.9: 4697180-4697180 on Build GRCh38
rs746014245 Chr.9: 4697208-4697208 on Build GRCh38
rs753698537 Chr.9: 4697047-4697047 on Build GRCh38
rs772713277 Chr.9: 4697138-4697138 on Build GRCh38
rs747082843 Chr.9: 4697073-4697073 on Build GRCh38
rs749731003 Chr.9: 4697116-4697116 on Build GRCh38
rs370371135 Chr.9: 4697048-4697048 on Build GRCh38
rs777908567 Chr.9: 4697061-4697061 on Build GRCh38
rs888522205 Chr.9: 4697083-4697084 on Build GRCh38
rs540369719 Chr.9: 4697172-4697172 on Build GRCh38
rs760600624 Chr.9: 4697139-4697139 on Build GRCh38
rs373184488 Chr.9: 4697079-4697079 on Build GRCh38
rs758631249 Chr.9: 4697059-4697059 on Build GRCh38
rs765112410 Chr.9: 4697154-4697154 on Build GRCh38
rs766359724 Chr.9: 4697145-4697145 on Build GRCh38
rs1040199525 Chr.9: 4697096-4697096 on Build GRCh38
rs754771656 Chr.9: 4697151-4697151 on Build GRCh38
rs200458324 Chr.9: 4697203-4697203 on Build GRCh38
rs146272968 Chr.9: 4697149-4697149 on Build GRCh38
rs773831988 Chr.9: 4697121-4697121 on Build GRCh38
rs7025425 Chr.9: 4697080-4697080 on Build GRCh38
rs367867848 Chr.9: 4697123-4697123 on Build GRCh38

View Full Product Details