Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 236580620-236580865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACKR3
Entrez Gene ID
57007
Gene Name
atypical chemokine receptor 3
Gene Aliases
CMKOR1, CXC-R7, CXCR-7, CXCR7, GPR159, RDC-1, RDC1
UniGene
Hs.471751
Gene Chromosome Location
Chr.2: 236537131-236582358 on Build GRCh38
Transcripts
NM_020311.2 XM_005246097.2 XM_005246098.3 XM_017004516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6592124 c.194T>C p.V65A Chr.2: 236580659-236580659 on Build GRCh38
1531337 c.226G>A p.G76S Chr.2: 236580691-236580691 on Build GRCh38
4092621 c.348G>A p.T116T Chr.2: 236580813-236580813 on Build GRCh38
5699839 c.289A>T p.I97F Chr.2: 236580754-236580754 on Build GRCh38
3579561 c.165C>T p.F55F Chr.2: 236580630-236580630 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779529043 Chr.2: 236580831-236580831 on Build GRCh38
rs368363375 Chr.2: 236580702-236580702 on Build GRCh38
rs145205829 Chr.2: 236580736-236580736 on Build GRCh38
rs202036823 Chr.2: 236580726-236580726 on Build GRCh38
rs765502728 Chr.2: 236580667-236580667 on Build GRCh38
rs777648022 Chr.2: 236580698-236580698 on Build GRCh38
rs762709549 Chr.2: 236580804-236580804 on Build GRCh38
rs114269199 Chr.2: 236580655-236580655 on Build GRCh38
rs149094897 Chr.2: 236580777-236580777 on Build GRCh38
rs569198092 Chr.2: 236580664-236580664 on Build GRCh38
rs773823324 Chr.2: 236580754-236580754 on Build GRCh38
rs573798270 Chr.2: 236580716-236580716 on Build GRCh38
rs772591745 Chr.2: 236580738-236580738 on Build GRCh38
rs761313182 Chr.2: 236580755-236580755 on Build GRCh38
rs763930150 Chr.2: 236580805-236580805 on Build GRCh38
rs1042377965 Chr.2: 236580819-236580819 on Build GRCh38
rs528077974 Chr.2: 236580813-236580813 on Build GRCh38
rs142866883 Chr.2: 236580778-236580778 on Build GRCh38
rs200095631 Chr.2: 236580701-236580701 on Build GRCh38
rs752378876 Chr.2: 236580678-236580678 on Build GRCh38
rs147420191 Chr.2: 236580732-236580732 on Build GRCh38
rs771119606 Chr.2: 236580762-236580762 on Build GRCh38
rs752994969 Chr.2: 236580802-236580802 on Build GRCh38
rs144999866 Chr.2: 236580812-236580812 on Build GRCh38
rs765328546 Chr.2: 236580789-236580789 on Build GRCh38
rs773878214 Chr.2: 236580651-236580651 on Build GRCh38
rs35767135 Chr.2: 236580654-236580654 on Build GRCh38
rs200840916 Chr.2: 236580695-236580695 on Build GRCh38
rs748988996 Chr.2: 236580837-236580837 on Build GRCh38

View Full Product Details