Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 132752386-132752617 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VNN2
Entrez Gene ID
8875
Gene Name
vanin 2
Gene Aliases
FOAP-4, GPI-80
UniGene
Hs.293130
Gene Chromosome Location
Chr.6: 132743503-132763489 on Build GRCh38
Transcripts
NM_001242350.2 NM_004665.4 NM_078488.2 NR_034173.2 NR_034174.2 NR_110143.1 NR_110144.1 NR_110145.1 NR_110146.1 XM_006715593.3 XM_011536231.1 XM_017011407.1 XM_017011408.1 XM_017011409.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5671413 c.768G>T p.M256I Chr.6: 132752519-132752519 on Build GRCh38
5065042 c.757G>A p.A253T Chr.6: 132752530-132752530 on Build GRCh38
373250 c.705C>A p.P235P Chr.6: 132752582-132752582 on Build GRCh38
6105121 c.731T>C p.L244P Chr.6: 132752556-132752556 on Build GRCh38
1073447 c.800C>T p.T267I Chr.6: 132752487-132752487 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560768824 Chr.6: 132752514-132752514 on Build GRCh38
rs183534398 Chr.6: 132752429-132752429 on Build GRCh38
rs755046321 Chr.6: 132752571-132752571 on Build GRCh38
rs768908410 Chr.6: 132752466-132752467 on Build GRCh38
rs758913232 Chr.6: 132752435-132752435 on Build GRCh38
rs201713695 Chr.6: 132752459-132752459 on Build GRCh38
rs375470188 Chr.6: 132752547-132752547 on Build GRCh38
rs1035238889 Chr.6: 132752428-132752428 on Build GRCh38
rs756347080 Chr.6: 132752584-132752584 on Build GRCh38
rs368725042 Chr.6: 132752512-132752512 on Build GRCh38
rs781246991 Chr.6: 132752460-132752460 on Build GRCh38
rs770854489 Chr.6: 132752505-132752505 on Build GRCh38
rs538255742 Chr.6: 132752415-132752415 on Build GRCh38
rs372326341 Chr.6: 132752524-132752524 on Build GRCh38
rs143802266 Chr.6: 132752523-132752523 on Build GRCh38
rs544259379 Chr.6: 132752499-132752499 on Build GRCh38
rs780338538 Chr.6: 132752441-132752441 on Build GRCh38
rs774361143 Chr.6: 132752550-132752550 on Build GRCh38
rs777615410 Chr.6: 132752593-132752593 on Build GRCh38
rs33920182 Chr.6: 132752566-132752566 on Build GRCh38
rs748332497 Chr.6: 132752581-132752581 on Build GRCh38
rs765594397 Chr.6: 132752420-132752420 on Build GRCh38
rs962459557 Chr.6: 132752587-132752587 on Build GRCh38
rs760306479 Chr.6: 132752521-132752521 on Build GRCh38
rs772934935 Chr.6: 132752491-132752491 on Build GRCh38
rs748364026 Chr.6: 132752462-132752462 on Build GRCh38
rs781268124 Chr.6: 132752579-132752579 on Build GRCh38
rs956224237 Chr.6: 132752481-132752481 on Build GRCh38
rs769483048 Chr.6: 132752485-132752485 on Build GRCh38
rs755266361 Chr.6: 132752456-132752456 on Build GRCh38
rs775034677 Chr.6: 132752517-132752517 on Build GRCh38
rs747080103 Chr.6: 132752452-132752452 on Build GRCh38
rs773894418 Chr.6: 132752511-132752511 on Build GRCh38
rs749055674 Chr.6: 132752594-132752594 on Build GRCh38

View Full Product Details