Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121378837-121379072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3C
Entrez Gene ID
10447
Gene Name
family with sequence similarity 3 member C
Gene Aliases
GS3786, ILEI
UniGene
Hs.434053
Gene Chromosome Location
Chr.7: 121348851-121396369 on Build GRCh38
Transcripts
NM_001040020.1 NM_014888.2 XM_011515736.2 XM_011515737.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
452217 c.117T>G p.F39L Chr.7: 121378911-121378911 on Build GRCh38
1084591 c.88A>C p.K30Q Chr.7: 121378940-121378940 on Build GRCh38
5881310 c.73C>A p.Q25K Chr.7: 121378955-121378955 on Build GRCh38
7102988 c.28G>A p.V10M Chr.7: 121379000-121379000 on Build GRCh38
5629618 c.91A>G p.M31V Chr.7: 121378937-121378937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs151288890 Chr.7: 121378934-121378934 on Build GRCh38
rs923673597 Chr.7: 121378890-121378890 on Build GRCh38
rs749522040 Chr.7: 121378933-121378933 on Build GRCh38
rs948973252 Chr.7: 121379000-121379000 on Build GRCh38
rs768266059 Chr.7: 121378914-121378914 on Build GRCh38
rs747465982 Chr.7: 121379012-121379012 on Build GRCh38
rs752403094 Chr.7: 121378885-121378885 on Build GRCh38
rs769442838 Chr.7: 121379025-121379025 on Build GRCh38
rs773826328 Chr.7: 121378985-121378985 on Build GRCh38
rs748584682 Chr.7: 121378972-121378972 on Build GRCh38
rs904882855 Chr.7: 121379037-121379037 on Build GRCh38
rs1044739712 Chr.7: 121379035-121379035 on Build GRCh38
rs764004885 Chr.7: 121378910-121378910 on Build GRCh38
rs762727321 Chr.7: 121379045-121379045 on Build GRCh38
rs372222980 Chr.7: 121378882-121378882 on Build GRCh38
rs750318600 Chr.7: 121378917-121378917 on Build GRCh38
rs745907404 Chr.7: 121378930-121378930 on Build GRCh38
rs759133488 Chr.7: 121378870-121378870 on Build GRCh38
rs564048822 Chr.7: 121378935-121378935 on Build GRCh38
rs756974338 Chr.7: 121378913-121378913 on Build GRCh38
rs751362515 Chr.7: 121378911-121378911 on Build GRCh38
rs376952658 Chr.7: 121378936-121378936 on Build GRCh38
rs780037177 Chr.7: 121378931-121378931 on Build GRCh38
rs772300135 Chr.7: 121378975-121378975 on Build GRCh38
rs756287917 Chr.7: 121378927-121378927 on Build GRCh38
rs762637223 Chr.7: 121378904-121378904 on Build GRCh38
rs936434467 Chr.7: 121378899-121378899 on Build GRCh38
rs914992663 Chr.7: 121378926-121378926 on Build GRCh38
rs1054599048 Chr.7: 121378932-121378932 on Build GRCh38
rs775092417 Chr.7: 121379031-121379031 on Build GRCh38

View Full Product Details