Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 107950242-107950493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2U1
Entrez Gene ID
113612
Gene Name
cytochrome P450 family 2 subfamily U member 1
Gene Aliases
P450TEC, SPG49, SPG56
UniGene
Hs.109087
Gene Chromosome Location
Chr.4: 107931549-107953461 on Build GRCh38
Transcripts
NM_183075.2 XM_005262717.2 XM_005262720.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4121217 c.1570C>A p.L524M Chr.4: 107950358-107950358 on Build GRCh38
4851115 c.1634G>T p.*545L Chr.4: 107950422-107950422 on Build GRCh38
5014444 c.1497A>C p.E499D Chr.4: 107950285-107950285 on Build GRCh38
5681317 c.1603C>T p.H535Y Chr.4: 107950391-107950391 on Build GRCh38
4527564 c.1473G>T p.M491I Chr.4: 107950261-107950261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772136947 Chr.4: 107950418-107950418 on Build GRCh38
rs576923939 Chr.4: 107950419-107950419 on Build GRCh38
rs752917310 Chr.4: 107950281-107950281 on Build GRCh38
rs760935150 Chr.4: 107950424-107950424 on Build GRCh38
rs762225242 Chr.4: 107950438-107950438 on Build GRCh38
rs977349364 Chr.4: 107950421-107950421 on Build GRCh38
rs773047007 Chr.4: 107950372-107950372 on Build GRCh38
rs780578327 Chr.4: 107950336-107950336 on Build GRCh38
rs79009744 Chr.4: 107950454-107950454 on Build GRCh38
rs770825730 Chr.4: 107950393-107950393 on Build GRCh38
rs144144031 Chr.4: 107950403-107950403 on Build GRCh38
rs370914955 Chr.4: 107950444-107950444 on Build GRCh38
rs995234654 Chr.4: 107950307-107950307 on Build GRCh38
rs749132333 Chr.4: 107950376-107950376 on Build GRCh38
rs754308246 Chr.4: 107950312-107950312 on Build GRCh38
rs538538497 Chr.4: 107950442-107950442 on Build GRCh38
rs201919750 Chr.4: 107950434-107950434 on Build GRCh38
rs1028966851 Chr.4: 107950313-107950313 on Build GRCh38
rs139224667 Chr.4: 107950324-107950324 on Build GRCh38
rs377467252 Chr.4: 107950427-107950427 on Build GRCh38
rs373549777 Chr.4: 107950360-107950360 on Build GRCh38
rs183327899 Chr.4: 107950457-107950457 on Build GRCh38
rs765717394 Chr.4: 107950443-107950443 on Build GRCh38
rs1033325675 Chr.4: 107950435-107950435 on Build GRCh38
rs1036607521 Chr.4: 107950269-107950269 on Build GRCh38
rs371017677 Chr.4: 107950458-107950458 on Build GRCh38
rs529806409 Chr.4: 107950433-107950433 on Build GRCh38
rs759494725 Chr.4: 107950412-107950412 on Build GRCh38
rs150818323 Chr.4: 107950322-107950322 on Build GRCh38
rs756308496 Chr.4: 107950295-107950295 on Build GRCh38
rs372600671 Chr.4: 107950321-107950321 on Build GRCh38
rs747768954 Chr.4: 107950352-107950352 on Build GRCh38
rs759654099 Chr.4: 107950271-107950271 on Build GRCh38
rs764456836 Chr.4: 107950297-107950297 on Build GRCh38
rs754487808 Chr.4: 107950334-107950334 on Build GRCh38
rs764362029 Chr.4: 107950314-107950314 on Build GRCh38
rs780269996 Chr.4: 107950452-107950453 on Build GRCh38
rs368543204 Chr.4: 107950280-107950280 on Build GRCh38

View Full Product Details