Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 5923305-5923530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHGB
Entrez Gene ID
1114
Gene Name
chromogranin B
Gene Aliases
SCG1
UniGene
Hs.516874
Gene Chromosome Location
Chr.20: 5911328-5925361 on Build GRCh38
Transcripts
NM_001819.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
149135 c.1250G>A p.R417H Chr.20: 5923394-5923394 on Build GRCh38
5930006 c.1169C>T p.P390L Chr.20: 5923313-5923313 on Build GRCh38
7323819 c.1309G>C p.E437Q Chr.20: 5923453-5923453 on Build GRCh38
6628357 c.1239G>A p.P413P Chr.20: 5923383-5923383 on Build GRCh38
7361177 c.1259G>A p.R420K Chr.20: 5923403-5923403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768902329 Chr.20: 5923328-5923328 on Build GRCh38
rs765892874 Chr.20: 5923373-5923373 on Build GRCh38
rs780332142 Chr.20: 5923418-5923418 on Build GRCh38
rs267606033 Chr.20: 5923434-5923434 on Build GRCh38
rs742711 Chr.20: 5923394-5923394 on Build GRCh38
rs748250834 Chr.20: 5923443-5923443 on Build GRCh38
rs746929287 Chr.20: 5923358-5923358 on Build GRCh38
rs865861615 Chr.20: 5923432-5923432 on Build GRCh38
rs373817587 Chr.20: 5923393-5923393 on Build GRCh38
rs367665937 Chr.20: 5923419-5923419 on Build GRCh38
rs757302423 Chr.20: 5923509-5923509 on Build GRCh38
rs989694700 Chr.20: 5923338-5923338 on Build GRCh38
rs530644191 Chr.20: 5923409-5923409 on Build GRCh38
rs768816863 Chr.20: 5923422-5923422 on Build GRCh38
rs764003023 Chr.20: 5923484-5923484 on Build GRCh38
rs764598566 Chr.20: 5923478-5923478 on Build GRCh38
rs762473977 Chr.20: 5923480-5923480 on Build GRCh38
rs200027232 Chr.20: 5923349-5923349 on Build GRCh38
rs752402192 Chr.20: 5923479-5923479 on Build GRCh38
rs779208717 Chr.20: 5923423-5923423 on Build GRCh38
rs742710 Chr.20: 5923382-5923382 on Build GRCh38
rs911967267 Chr.20: 5923424-5923424 on Build GRCh38
rs756888615 Chr.20: 5923452-5923453 on Build GRCh38
rs200140204 Chr.20: 5923407-5923407 on Build GRCh38
rs772514466 Chr.20: 5923445-5923445 on Build GRCh38
rs749372552 Chr.20: 5923420-5923420 on Build GRCh38
rs868756086 Chr.20: 5923343-5923343 on Build GRCh38
rs781260806 Chr.20: 5923510-5923510 on Build GRCh38
rs761127764 Chr.20: 5923365-5923365 on Build GRCh38
rs771393741 Chr.20: 5923460-5923460 on Build GRCh38
rs74728352 Chr.20: 5923477-5923477 on Build GRCh38
rs773512156 Chr.20: 5923453-5923453 on Build GRCh38
rs193921088 Chr.20: 5923452-5923452 on Build GRCh38
rs774644338 Chr.20: 5923333-5923333 on Build GRCh38
rs753321054 Chr.20: 5923378-5923378 on Build GRCh38
rs759076072 Chr.20: 5923465-5923465 on Build GRCh38
rs918538290 Chr.20: 5923483-5923483 on Build GRCh38
rs757214500 Chr.20: 5923412-5923412 on Build GRCh38
rs760016873 Chr.20: 5923500-5923500 on Build GRCh38
rs371474057 Chr.20: 5923507-5923507 on Build GRCh38

View Full Product Details