Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 49491146-49491419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF423
Entrez Gene ID
23090
Gene Name
zinc finger protein 423
Gene Aliases
Ebfaz, JBTS19, NPHP14, OAZ, Roaz, ZFP423, Zfp104, hOAZ
UniGene
Hs.443715
Gene Chromosome Location
Chr.16: 49487531-49857919 on Build GRCh38
Transcripts
NM_001271620.2 NM_015069.4 XM_005255856.4 XM_005255857.4 XM_006721171.3 XM_017023076.1 XM_017023077.1 XM_017023078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
362892 c.3833C>T p.T1278M Chr.16: 49491297-49491297 on Build GRCh38
5985076 c.3837G>C p.M1279I Chr.16: 49491293-49491293 on Build GRCh38
6311232 c.3846C>A p.H1282Q Chr.16: 49491284-49491284 on Build GRCh38
362891 c.3833C>T p.T1278M Chr.16: 49491297-49491297 on Build GRCh38
5985077 c.3837G>C p.M1279I Chr.16: 49491293-49491293 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764865276 Chr.16: 49491204-49491204 on Build GRCh38
rs202192192 Chr.16: 49491248-49491248 on Build GRCh38
rs751807499 Chr.16: 49491252-49491253 on Build GRCh38
rs544552211 Chr.16: 49491290-49491290 on Build GRCh38
rs914716751 Chr.16: 49491336-49491338 on Build GRCh38
rs947895961 Chr.16: 49491359-49491359 on Build GRCh38
rs896823232 Chr.16: 49491394-49491394 on Build GRCh38
rs777569372 Chr.16: 49491319-49491319 on Build GRCh38
rs770872541 Chr.16: 49491241-49491241 on Build GRCh38
rs200788408 Chr.16: 49491338-49491338 on Build GRCh38
rs749467480 Chr.16: 49491343-49491343 on Build GRCh38
rs139544621 Chr.16: 49491229-49491229 on Build GRCh38
rs769972969 Chr.16: 49491261-49491261 on Build GRCh38
rs948904725 Chr.16: 49491340-49491340 on Build GRCh38
rs953862247 Chr.16: 49491185-49491185 on Build GRCh38
rs927679086 Chr.16: 49491392-49491392 on Build GRCh38
rs774772870 Chr.16: 49491355-49491355 on Build GRCh38
rs763108153 Chr.16: 49491278-49491278 on Build GRCh38
rs1036735609 Chr.16: 49491385-49491385 on Build GRCh38
rs559197075 Chr.16: 49491268-49491268 on Build GRCh38
rs530104828 Chr.16: 49491325-49491325 on Build GRCh38
rs369770026 Chr.16: 49491234-49491234 on Build GRCh38
rs12445683 Chr.16: 49491327-49491327 on Build GRCh38
rs751372572 Chr.16: 49491321-49491321 on Build GRCh38
rs369646519 Chr.16: 49491297-49491297 on Build GRCh38
rs78514142 Chr.16: 49491313-49491313 on Build GRCh38
rs145999684 Chr.16: 49491178-49491178 on Build GRCh38
rs771300748 Chr.16: 49491225-49491225 on Build GRCh38
rs775616540 Chr.16: 49491307-49491307 on Build GRCh38
rs373946136 Chr.16: 49491192-49491192 on Build GRCh38
rs757954584 Chr.16: 49491386-49491386 on Build GRCh38
rs879213062 Chr.16: 49491176-49491176 on Build GRCh38
rs907478029 Chr.16: 49491360-49491360 on Build GRCh38
rs776683301 Chr.16: 49491242-49491242 on Build GRCh38
rs755123810 Chr.16: 49491357-49491357 on Build GRCh38
rs150302551 Chr.16: 49491279-49491279 on Build GRCh38
rs745926866 Chr.16: 49491329-49491329 on Build GRCh38
rs1007586630 Chr.16: 49491228-49491228 on Build GRCh38
rs775831924 Chr.16: 49491270-49491270 on Build GRCh38
rs779999900 Chr.16: 49491339-49491339 on Build GRCh38
rs374808247 Chr.16: 49491233-49491233 on Build GRCh38
rs938345608 Chr.16: 49491384-49491384 on Build GRCh38
rs768817161 Chr.16: 49491346-49491346 on Build GRCh38
rs774591168 Chr.16: 49491282-49491282 on Build GRCh38
rs759162963 Chr.16: 49491296-49491296 on Build GRCh38
rs976147381 Chr.16: 49491264-49491264 on Build GRCh38
rs768204840 Chr.16: 49491287-49491287 on Build GRCh38
rs573236278 Chr.16: 49491226-49491226 on Build GRCh38
rs762394882 Chr.16: 49491284-49491284 on Build GRCh38
rs746900694 Chr.16: 49491235-49491235 on Build GRCh38
rs748497366 Chr.16: 49491378-49491379 on Build GRCh38
rs746015413 Chr.16: 49491253-49491253 on Build GRCh38

View Full Product Details