Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19134185-19134448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGCR14
Entrez Gene ID
8220
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
DGCR13, DGS-H, DGS-I, DGSH, DGSI, ES2, Es2el
UniGene
Hs.517407
Gene Chromosome Location
Chr.22: 19130279-19144726 on Build GRCh38
Transcripts
NM_022719.2 NR_134304.1 XM_005261282.4 XM_006724329.3 XM_006724330.3 XM_006724331.3 XM_017028958.1 XM_017028959.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3842171 c.1305G>T p.L435L Chr.22: 19134322-19134322 on Build GRCh38
1484069 c.1293G>A p.P431P Chr.22: 19134334-19134334 on Build GRCh38
5393143 c.1425C>A p.F475L Chr.22: 19134202-19134202 on Build GRCh38
1714140 c.1313C>T p.P438L Chr.22: 19134314-19134314 on Build GRCh38
6642858 c.1246G>A p.A416T Chr.22: 19134381-19134381 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776531141 Chr.22: 19134356-19134356 on Build GRCh38
rs780144981 Chr.22: 19134299-19134299 on Build GRCh38
rs751161380 Chr.22: 19134327-19134327 on Build GRCh38
rs186717850 Chr.22: 19134393-19134393 on Build GRCh38
rs762542001 Chr.22: 19134318-19134318 on Build GRCh38
rs575074091 Chr.22: 19134428-19134428 on Build GRCh38
rs768922267 Chr.22: 19134302-19134302 on Build GRCh38
rs367552506 Chr.22: 19134285-19134285 on Build GRCh38
rs768834089 Chr.22: 19134253-19134253 on Build GRCh38
rs757580968 Chr.22: 19134328-19134328 on Build GRCh38
rs761563805 Chr.22: 19134396-19134396 on Build GRCh38
rs777185987 Chr.22: 19134219-19134219 on Build GRCh38
rs200592269 Chr.22: 19134350-19134350 on Build GRCh38
rs749362875 Chr.22: 19134301-19134301 on Build GRCh38
rs373680781 Chr.22: 19134247-19134247 on Build GRCh38
rs749383026 Chr.22: 19134345-19134345 on Build GRCh38
rs138868865 Chr.22: 19134266-19134266 on Build GRCh38
rs712965 Chr.22: 19134359-19134359 on Build GRCh38
rs751194608 Chr.22: 19134357-19134357 on Build GRCh38
rs763671482 Chr.22: 19134324-19134324 on Build GRCh38
rs551191358 Chr.22: 19134282-19134282 on Build GRCh38
rs748146654 Chr.22: 19134429-19134429 on Build GRCh38
rs746322500 Chr.22: 19134297-19134297 on Build GRCh38
rs766455458 Chr.22: 19134394-19134395 on Build GRCh38
rs781653916 Chr.22: 19134333-19134333 on Build GRCh38
rs766241326 Chr.22: 19134260-19134260 on Build GRCh38
rs113582836 Chr.22: 19134242-19134242 on Build GRCh38
rs533285660 Chr.22: 19134314-19134314 on Build GRCh38
rs772946229 Chr.22: 19134258-19134258 on Build GRCh38
rs766875280 Chr.22: 19134410-19134410 on Build GRCh38
rs1045333771 Chr.22: 19134235-19134235 on Build GRCh38
rs745952527 Chr.22: 19134221-19134221 on Build GRCh38
rs539108063 Chr.22: 19134334-19134334 on Build GRCh38
rs769912652 Chr.22: 19134369-19134369 on Build GRCh38
rs750843358 Chr.22: 19134404-19134404 on Build GRCh38
rs146964365 Chr.22: 19134309-19134309 on Build GRCh38
rs747371131 Chr.22: 19134213-19134213 on Build GRCh38
rs770720252 Chr.22: 19134310-19134310 on Build GRCh38
rs377733215 Chr.22: 19134294-19134294 on Build GRCh38
rs767141147 Chr.22: 19134409-19134409 on Build GRCh38
rs200628413 Chr.22: 19134218-19134218 on Build GRCh38
rs1036269035 Chr.22: 19134332-19134332 on Build GRCh38
rs142803658 Chr.22: 19134281-19134281 on Build GRCh38
rs368666308 Chr.22: 19134298-19134298 on Build GRCh38
rs764793591 Chr.22: 19134268-19134268 on Build GRCh38
rs1052891938 Chr.22: 19134388-19134388 on Build GRCh38
rs975182762 Chr.22: 19134291-19134291 on Build GRCh38
rs778713377 Chr.22: 19134207-19134207 on Build GRCh38
rs774300373 Chr.22: 19134337-19134337 on Build GRCh38
rs763606714 Chr.22: 19134392-19134392 on Build GRCh38
rs137995273 Chr.22: 19134338-19134338 on Build GRCh38
rs765237389 Chr.22: 19134317-19134317 on Build GRCh38
rs767070613 Chr.22: 19134403-19134403 on Build GRCh38
rs377464869 Chr.22: 19134384-19134384 on Build GRCh38
rs768714246 Chr.22: 19134346-19134346 on Build GRCh38
rs962807666 Chr.22: 19134290-19134290 on Build GRCh38
rs143848509 Chr.22: 19134415-19134415 on Build GRCh38
rs760504940 Chr.22: 19134259-19134259 on Build GRCh38
rs991861629 Chr.22: 19134342-19134342 on Build GRCh38
rs757587876 Chr.22: 19134286-19134286 on Build GRCh38
rs61737477 Chr.22: 19134209-19134209 on Build GRCh38
rs905020907 Chr.22: 19134244-19134244 on Build GRCh38
rs775514047 Chr.22: 19134383-19134383 on Build GRCh38
rs373227576 Chr.22: 19134349-19134349 on Build GRCh38
rs776455586 Chr.22: 19134313-19134313 on Build GRCh38
rs753742890 Chr.22: 19134261-19134261 on Build GRCh38
rs375484595 Chr.22: 19134222-19134222 on Build GRCh38
rs373320979 Chr.22: 19134289-19134289 on Build GRCh38
rs929282 Chr.22: 19134208-19134208 on Build GRCh38
rs779035330 Chr.22: 19134348-19134348 on Build GRCh38
rs754302679 Chr.22: 19134412-19134412 on Build GRCh38
rs534156960 Chr.22: 19134431-19134431 on Build GRCh38
rs756678975 Chr.22: 19134335-19134335 on Build GRCh38
rs369841251 Chr.22: 19134241-19134241 on Build GRCh38
rs769820225 Chr.22: 19134234-19134234 on Build GRCh38
rs368459829 Chr.22: 19134306-19134306 on Build GRCh38

View Full Product Details