Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 33291202-33291461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC2
Entrez Gene ID
40
Gene Name
acid sensing ion channel subunit 2
Gene Aliases
ACCN, ACCN1, ASIC2a, BNC1, BNaC1, MDEG, hBNaC1
UniGene
Hs.368417
Gene Chromosome Location
Chr.17: 33013087-34156806 on Build GRCh38
Transcripts
NM_001094.4 NM_183377.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5426881 c.708+9C>A p.? Chr.17: 33291399-33291399 on Build GRCh38
6417554 c.691C>G p.P231A Chr.17: 33291425-33291425 on Build GRCh38
1610068 c.676G>T p.G226C Chr.17: 33291440-33291440 on Build GRCh38
6605745 c.693G>A p.P231P Chr.17: 33291423-33291423 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757977999 Chr.17: 33291413-33291415 on Build GRCh38
rs749626842 Chr.17: 33291432-33291432 on Build GRCh38
rs772266671 Chr.17: 33291224-33291224 on Build GRCh38
rs752537589 Chr.17: 33291383-33291383 on Build GRCh38
rs775025267 Chr.17: 33291361-33291361 on Build GRCh38
rs1027700901 Chr.17: 33291428-33291428 on Build GRCh38
rs141720961 Chr.17: 33291426-33291426 on Build GRCh38
rs185077500 Chr.17: 33291377-33291377 on Build GRCh38
rs767652881 Chr.17: 33291371-33291371 on Build GRCh38
rs536066627 Chr.17: 33291289-33291289 on Build GRCh38
rs570328092 Chr.17: 33291263-33291263 on Build GRCh38
rs761735967 Chr.17: 33291364-33291364 on Build GRCh38
rs756706831 Chr.17: 33291408-33291408 on Build GRCh38
rs896440209 Chr.17: 33291368-33291368 on Build GRCh38
rs549261422 Chr.17: 33291319-33291319 on Build GRCh38
rs945158614 Chr.17: 33291280-33291280 on Build GRCh38
rs999269115 Chr.17: 33291356-33291356 on Build GRCh38
rs937656513 Chr.17: 33291341-33291341 on Build GRCh38
rs769532040 Chr.17: 33291425-33291425 on Build GRCh38
rs533640875 Chr.17: 33291390-33291390 on Build GRCh38
rs996470385 Chr.17: 33291232-33291232 on Build GRCh38
rs764988393 Chr.17: 33291381-33291381 on Build GRCh38
rs758213506 Chr.17: 33291384-33291384 on Build GRCh38
rs780819929 Chr.17: 33291412-33291412 on Build GRCh38
rs777619633 Chr.17: 33291389-33291389 on Build GRCh38
rs750600354 Chr.17: 33291373-33291373 on Build GRCh38
rs890668443 Chr.17: 33291293-33291293 on Build GRCh38
rs376011881 Chr.17: 33291391-33291391 on Build GRCh38
rs914055436 Chr.17: 33291265-33291265 on Build GRCh38
rs1055189992 Chr.17: 33291360-33291360 on Build GRCh38
rs1030897641 Chr.17: 33291239-33291239 on Build GRCh38
rs371466838 Chr.17: 33291226-33291226 on Build GRCh38
rs745476399 Chr.17: 33291423-33291423 on Build GRCh38
rs952150268 Chr.17: 33291438-33291438 on Build GRCh38

View Full Product Details