Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 102331765-102331990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF839
Entrez Gene ID
55778
Gene Name
zinc finger protein 839
Gene Aliases
C14orf131
UniGene
Hs.743359
Gene Chromosome Location
Chr.14: 102317377-102343174 on Build GRCh38
Transcripts
NM_001267827.1 NM_001267828.1 NM_018335.4 XM_006720203.3 XM_011536945.1 XM_011536946.2 XM_011536948.2 XM_011536949.2 XM_011536950.2 XM_017021447.1 XM_017021448.1 XM_017021449.1 XM_017021450.1 XM_017021451.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4825477 c.1391C>T p.S464L Chr.14: 102331821-102331821 on Build GRCh38
3419638 c.1379A>T p.E460V Chr.14: 102331809-102331809 on Build GRCh38
953537 c.1392G>A p.S464S Chr.14: 102331822-102331822 on Build GRCh38
4886026 c.1361G>T p.G454V Chr.14: 102331791-102331791 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750037013 Chr.14: 102331840-102331840 on Build GRCh38
rs773307456 Chr.14: 102331801-102331801 on Build GRCh38
rs960378496 Chr.14: 102331821-102331821 on Build GRCh38
rs1032787359 Chr.14: 102331818-102331818 on Build GRCh38
rs373962634 Chr.14: 102331800-102331800 on Build GRCh38
rs963989257 Chr.14: 102331863-102331863 on Build GRCh38
rs8014122 Chr.14: 102331871-102331871 on Build GRCh38
rs763623809 Chr.14: 102331832-102331832 on Build GRCh38
rs183279161 Chr.14: 102331950-102331950 on Build GRCh38
rs147106635 Chr.14: 102331879-102331879 on Build GRCh38
rs200235026 Chr.14: 102331819-102331819 on Build GRCh38
rs867389745 Chr.14: 102331785-102331785 on Build GRCh38
rs780952823 Chr.14: 102331838-102331838 on Build GRCh38
rs569344347 Chr.14: 102331833-102331833 on Build GRCh38
rs770912186 Chr.14: 102331882-102331882 on Build GRCh38
rs768651936 Chr.14: 102331874-102331874 on Build GRCh38
rs200326072 Chr.14: 102331888-102331888 on Build GRCh38
rs775228755 Chr.14: 102331880-102331880 on Build GRCh38
rs776954873 Chr.14: 102331891-102331891 on Build GRCh38
rs375163932 Chr.14: 102331837-102331837 on Build GRCh38
rs747001405 Chr.14: 102331909-102331909 on Build GRCh38
rs749386189 Chr.14: 102331841-102331841 on Build GRCh38
rs917700276 Chr.14: 102331959-102331959 on Build GRCh38
rs539059831 Chr.14: 102331810-102331810 on Build GRCh38
rs756455606 Chr.14: 102331932-102331932 on Build GRCh38
rs566852704 Chr.14: 102331804-102331804 on Build GRCh38
rs1054809839 Chr.14: 102331881-102331881 on Build GRCh38
rs368513189 Chr.14: 102331822-102331822 on Build GRCh38
rs952676387 Chr.14: 102331903-102331903 on Build GRCh38
rs765553277 Chr.14: 102331817-102331817 on Build GRCh38
rs746066533 Chr.14: 102331893-102331893 on Build GRCh38
rs541336680 Chr.14: 102331963-102331963 on Build GRCh38
rs374586476 Chr.14: 102331873-102331873 on Build GRCh38
rs770786436 Chr.14: 102331835-102331836 on Build GRCh38
rs756024755 Chr.14: 102331854-102331854 on Build GRCh38

View Full Product Details