Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9961001-9961254 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL1
Entrez Gene ID
29841
Gene Name
grainyhead like transcription factor 1
Gene Aliases
LBP32, MGR, NH32, TFCP2L2
UniGene
Hs.418493
Gene Chromosome Location
Chr.2: 9951663-10002284 on Build GRCh38
Transcripts
NM_198182.2 XM_005246159.3 XM_006711882.2 XM_006711884.3 XM_011510343.2 XM_017003900.1 XM_017003901.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4688453 c.452C>T p.T151M Chr.2: 9961219-9961219 on Build GRCh38
4688454 c.452C>T p.T151M Chr.2: 9961219-9961219 on Build GRCh38
6574597 c.315C>G p.I105M Chr.2: 9961082-9961082 on Build GRCh38
3564275 c.369C>T p.V123V Chr.2: 9961136-9961136 on Build GRCh38
6452384 c.428C>A p.T143K Chr.2: 9961195-9961195 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754495603 Chr.2: 9961160-9961160 on Build GRCh38
rs370641787 Chr.2: 9961150-9961150 on Build GRCh38
rs756658439 Chr.2: 9961196-9961196 on Build GRCh38
rs759674440 Chr.2: 9961071-9961071 on Build GRCh38
rs573056848 Chr.2: 9961063-9961063 on Build GRCh38
rs756983396 Chr.2: 9961195-9961195 on Build GRCh38
rs767616300 Chr.2: 9961081-9961081 on Build GRCh38
rs769040863 Chr.2: 9961144-9961144 on Build GRCh38
rs778188592 Chr.2: 9961208-9961208 on Build GRCh38
rs753762274 Chr.2: 9961082-9961082 on Build GRCh38
rs1024921410 Chr.2: 9961059-9961059 on Build GRCh38
rs747569983 Chr.2: 9961195-9961195 on Build GRCh38
rs1027865603 Chr.2: 9961124-9961124 on Build GRCh38
rs762037569 Chr.2: 9961159-9961159 on Build GRCh38
rs771273253 Chr.2: 9961214-9961214 on Build GRCh38
rs377444151 Chr.2: 9961068-9961068 on Build GRCh38
rs1048636619 Chr.2: 9961060-9961060 on Build GRCh38
rs780719872 Chr.2: 9961192-9961192 on Build GRCh38
rs772146837 Chr.2: 9961229-9961229 on Build GRCh38
rs750254394 Chr.2: 9961133-9961133 on Build GRCh38
rs774608719 Chr.2: 9961217-9961217 on Build GRCh38
rs746062803 Chr.2: 9961219-9961219 on Build GRCh38
rs761844045 Chr.2: 9961084-9961084 on Build GRCh38
rs765032179 Chr.2: 9961117-9961117 on Build GRCh38

View Full Product Details