Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32816780-32817041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HLA-DOB
Entrez Gene ID
3112
Gene Name
major histocompatibility complex, class II, DO beta
Gene Aliases
DOB
UniGene
Hs.1802
Gene Chromosome Location
Chr.6: 32812763-32817048 on Build GRCh38
Transcripts
NM_002120.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3662425 c.11G>T p.G4V Chr.6: 32816941-32816941 on Build GRCh38
3662426 c.1944G>T p.W648C Chr.6: 32816941-32816941 on Build GRCh38
5870327 c.58G>A p.D20N Chr.6: 32816894-32816894 on Build GRCh38
7177590 c.1933-1G>A p.? Chr.6: 32816953-32816953 on Build GRCh38
5870328 c.1991G>A p.G664E Chr.6: 32816894-32816894 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs55988835 Chr.6: 32816813-32816813 on Build GRCh38
rs751653725 Chr.6: 32816907-32816907 on Build GRCh38
rs553921719 Chr.6: 32816849-32816849 on Build GRCh38
rs781428784 Chr.6: 32816933-32816933 on Build GRCh38
rs2071472 Chr.6: 32816843-32816843 on Build GRCh38
rs757426122 Chr.6: 32816930-32816930 on Build GRCh38
rs774137652 Chr.6: 32816869-32816869 on Build GRCh38
rs761430311 Chr.6: 32816991-32816991 on Build GRCh38
rs112887968 Chr.6: 32816898-32816898 on Build GRCh38
rs2071471 Chr.6: 32816868-32816868 on Build GRCh38
rs567290998 Chr.6: 32816885-32816885 on Build GRCh38
rs2071554 Chr.6: 32816899-32816899 on Build GRCh38
rs773037752 Chr.6: 32817003-32817003 on Build GRCh38
rs753872785 Chr.6: 32816895-32816895 on Build GRCh38
rs543275553 Chr.6: 32816886-32816886 on Build GRCh38
rs956348294 Chr.6: 32816970-32816970 on Build GRCh38
rs746190236 Chr.6: 32816940-32816940 on Build GRCh38
rs761487088 Chr.6: 32816874-32816874 on Build GRCh38
rs1011979502 Chr.6: 32816932-32816932 on Build GRCh38
rs777764420 Chr.6: 32816902-32816902 on Build GRCh38
rs770246200 Chr.6: 32816951-32816951 on Build GRCh38
rs767290583 Chr.6: 32816878-32816878 on Build GRCh38
rs770163710 Chr.6: 32816825-32816825 on Build GRCh38
rs1047690297 Chr.6: 32816807-32816807 on Build GRCh38
rs549419981 Chr.6: 32816964-32816964 on Build GRCh38
rs2071470 Chr.6: 32816976-32816976 on Build GRCh38
rs776926902 Chr.6: 32816811-32816811 on Build GRCh38
rs544096527 Chr.6: 32816961-32816961 on Build GRCh38
rs780408640 Chr.6: 32816955-32816955 on Build GRCh38
rs771810386 Chr.6: 32816992-32816992 on Build GRCh38
rs2071469 Chr.6: 32817006-32817006 on Build GRCh38
rs555199989 Chr.6: 32816887-32816887 on Build GRCh38

View Full Product Details