Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92609364-92609589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF11
Entrez Gene ID
3832
Gene Name
kinesin family member 11
Gene Aliases
EG5, HKSP, KNSL1, MCLMR, TRIP5
UniGene
Hs.8878
Gene Chromosome Location
Chr.10: 92593068-92655395 on Build GRCh38
Transcripts
NM_004523.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6325331 c.593G>A p.G198D Chr.10: 92609404-92609404 on Build GRCh38
921474 c.617A>G p.N206S Chr.10: 92609428-92609428 on Build GRCh38
5569423 c.607A>G p.T203A Chr.10: 92609418-92609418 on Build GRCh38
4618941 c.636A>T p.Q212H Chr.10: 92609447-92609447 on Build GRCh38
4695872 c.652G>A p.A218T Chr.10: 92609463-92609463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753561329 Chr.10: 92609558-92609558 on Build GRCh38
rs775467988 Chr.10: 92609528-92609528 on Build GRCh38
rs749452545 Chr.10: 92609486-92609486 on Build GRCh38
rs975343732 Chr.10: 92609551-92609551 on Build GRCh38
rs763687626 Chr.10: 92609546-92609546 on Build GRCh38
rs182150439 Chr.10: 92609467-92609467 on Build GRCh38
rs767404022 Chr.10: 92609522-92609522 on Build GRCh38
rs759542620 Chr.10: 92609501-92609501 on Build GRCh38
rs1032967752 Chr.10: 92609566-92609566 on Build GRCh38
rs1013691656 Chr.10: 92609455-92609455 on Build GRCh38
rs1019021271 Chr.10: 92609430-92609430 on Build GRCh38
rs556943927 Chr.10: 92609427-92609427 on Build GRCh38
rs377385042 Chr.10: 92609544-92609544 on Build GRCh38
rs748213759 Chr.10: 92609417-92609417 on Build GRCh38
rs773481246 Chr.10: 92609424-92609424 on Build GRCh38
rs145656661 Chr.10: 92609490-92609490 on Build GRCh38

View Full Product Details