Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75267854-75268099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIF4GD
Entrez Gene ID
57409
Gene Name
MIF4G domain containing
Gene Aliases
AD023, MIFD, SLIP1
UniGene
Hs.325631
Gene Chromosome Location
Chr.17: 75266228-75271292 on Build GRCh38
Transcripts
NM_001242498.1 NM_001242500.1 NM_001242501.1 NM_020679.3 XM_005257530.1 XM_005257532.4 XM_005257533.1 XM_011525051.2 XM_011525052.2 XM_011525053.2 XM_011525054.2 XM_011525055.2 XM_011525057.2 XM_017024888.1 XM_017024889.1 XM_017024890.1 XM_017024891.1 XM_017024892.1 XM_017024893.1 XM_017024894.1 XM_017024895.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6712782 c.294+8delG p.? Chr.17: 75268075-75268075 on Build GRCh38
1610754 c.294+1G>T p.? Chr.17: 75268082-75268082 on Build GRCh38
4449770 c.301_302delAG p.S101fs*1 Chr.17: 75267894-75267895 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs916964519 Chr.17: 75267923-75267923 on Build GRCh38
rs936943281 Chr.17: 75267959-75267959 on Build GRCh38
rs755265166 Chr.17: 75267873-75267874 on Build GRCh38
rs924119528 Chr.17: 75267948-75267948 on Build GRCh38
rs745472900 Chr.17: 75267941-75267941 on Build GRCh38
rs768622182 Chr.17: 75268039-75268039 on Build GRCh38
rs763216203 Chr.17: 75267876-75267876 on Build GRCh38
rs771798971 Chr.17: 75267946-75267946 on Build GRCh38
rs372043201 Chr.17: 75267875-75267875 on Build GRCh38
rs375297679 Chr.17: 75267949-75267949 on Build GRCh38
rs994253970 Chr.17: 75268045-75268045 on Build GRCh38
rs767874152 Chr.17: 75267906-75267906 on Build GRCh38
rs1053938639 Chr.17: 75267960-75267960 on Build GRCh38
rs145256099 Chr.17: 75267887-75267887 on Build GRCh38
rs779870336 Chr.17: 75267953-75267953 on Build GRCh38
rs879005297 Chr.17: 75267952-75267952 on Build GRCh38
rs763020738 Chr.17: 75268041-75268041 on Build GRCh38
rs748272473 Chr.17: 75268031-75268031 on Build GRCh38
rs73356372 Chr.17: 75267981-75267981 on Build GRCh38
rs756612545 Chr.17: 75267911-75267911 on Build GRCh38
rs765641121 Chr.17: 75267894-75267895 on Build GRCh38
rs372046043 Chr.17: 75267939-75267939 on Build GRCh38
rs773344465 Chr.17: 75268040-75268040 on Build GRCh38
rs112577980 Chr.17: 75267908-75267908 on Build GRCh38
rs889796965 Chr.17: 75267969-75267969 on Build GRCh38
rs774577850 Chr.17: 75268078-75268078 on Build GRCh38
rs199715972 Chr.17: 75267873-75267873 on Build GRCh38
rs368453838 Chr.17: 75267929-75267929 on Build GRCh38
rs369853135 Chr.17: 75268032-75268032 on Build GRCh38
rs762277397 Chr.17: 75267876-75267877 on Build GRCh38
rs372444588 Chr.17: 75268068-75268068 on Build GRCh38
rs750705474 Chr.17: 75267931-75267932 on Build GRCh38
rs958333195 Chr.17: 75267885-75267885 on Build GRCh38
rs1038692205 Chr.17: 75268076-75268076 on Build GRCh38
rs982131174 Chr.17: 75267969-75267970 on Build GRCh38
rs988939754 Chr.17: 75267992-75267992 on Build GRCh38
rs777207640 Chr.17: 75267938-75267938 on Build GRCh38
rs755348654 Chr.17: 75267900-75267900 on Build GRCh38
rs888265387 Chr.17: 75268058-75268058 on Build GRCh38
rs1025925239 Chr.17: 75268049-75268049 on Build GRCh38
rs73356374 Chr.17: 75268001-75268001 on Build GRCh38
rs928917096 Chr.17: 75267973-75267973 on Build GRCh38

View Full Product Details