Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75569139-75569410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POLK
Entrez Gene ID
51426
Gene Name
polymerase (DNA) kappa
Gene Aliases
DINB1, DINP, POLQ
UniGene
Hs.135756
Gene Chromosome Location
Chr.5: 75511832-75602576 on Build GRCh38
Transcripts
NM_016218.2 XM_005248534.4 XM_011543463.2 XM_011543464.2 XM_011543467.2 XM_017009558.1 XM_017009559.1 XM_017009560.1 XM_017009561.1 XM_017009562.1 XM_017009563.1 XM_017009564.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4844626 c.319G>A p.D107N Chr.5: 75569403-75569403 on Build GRCh38
4844624 c.319G>A p.D107N Chr.5: 75569403-75569403 on Build GRCh38
5567115 c.290G>A p.R97Q Chr.5: 75569374-75569374 on Build GRCh38
5115240 c.287G>T p.S96I Chr.5: 75569371-75569371 on Build GRCh38
3828431 c.304A>G p.T102A Chr.5: 75569388-75569388 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182952181 Chr.5: 75569177-75569177 on Build GRCh38
rs35011708 Chr.5: 75569226-75569227 on Build GRCh38
rs957145686 Chr.5: 75569320-75569320 on Build GRCh38
rs770423121 Chr.5: 75569373-75569373 on Build GRCh38
rs1040342754 Chr.5: 75569274-75569274 on Build GRCh38
rs944699507 Chr.5: 75569259-75569259 on Build GRCh38
rs373630071 Chr.5: 75569216-75569216 on Build GRCh38
rs755004522 Chr.5: 75569345-75569345 on Build GRCh38
rs757430614 Chr.5: 75569305-75569305 on Build GRCh38
rs772860689 Chr.5: 75569362-75569362 on Build GRCh38
rs1047027400 Chr.5: 75569208-75569208 on Build GRCh38
rs866995491 Chr.5: 75569266-75569266 on Build GRCh38
rs900039935 Chr.5: 75569252-75569252 on Build GRCh38
rs1469051 Chr.5: 75569224-75569224 on Build GRCh38
rs778173593 Chr.5: 75569295-75569295 on Build GRCh38
rs972243633 Chr.5: 75569204-75569205 on Build GRCh38
rs749180721 Chr.5: 75569328-75569328 on Build GRCh38
rs529866002 Chr.5: 75569329-75569329 on Build GRCh38
rs778538658 Chr.5: 75569348-75569348 on Build GRCh38
rs776261097 Chr.5: 75569384-75569384 on Build GRCh38
rs779978926 Chr.5: 75569313-75569313 on Build GRCh38
rs930007423 Chr.5: 75569204-75569204 on Build GRCh38
rs888534528 Chr.5: 75569211-75569211 on Build GRCh38
rs142907860 Chr.5: 75569352-75569352 on Build GRCh38
rs751641390 Chr.5: 75569299-75569299 on Build GRCh38
rs917135748 Chr.5: 75569174-75569174 on Build GRCh38
rs370103947 Chr.5: 75569372-75569372 on Build GRCh38
rs986363647 Chr.5: 75569248-75569248 on Build GRCh38
rs750145522 Chr.5: 75569325-75569325 on Build GRCh38
rs546760534 Chr.5: 75569355-75569355 on Build GRCh38

View Full Product Details