Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 41513960-41514217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT15
Entrez Gene ID
3866
Gene Name
keratin 15
Gene Aliases
CK15, K15, K1CO
UniGene
Hs.654570
Gene Chromosome Location
Chr.17: 41513745-41522413 on Build GRCh38
Transcripts
NM_002275.3 XM_011524784.2 XM_017024614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1750029 c.1333G>A p.D445N Chr.17: 41514061-41514061 on Build GRCh38
3517435 c.1332G>A p.V444V Chr.17: 41514062-41514062 on Build GRCh38
5555518 c.1288G>C p.G430R Chr.17: 41514106-41514106 on Build GRCh38
6580279 c.1315_1316insTCA p.I438_N439insI Chr.17: 41514078-41514079 on Build GRCh38
1383192 c.1363G>A p.E455K Chr.17: 41514031-41514031 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150390543 Chr.17: 41514062-41514062 on Build GRCh38
rs771422905 Chr.17: 41514097-41514097 on Build GRCh38
rs751654645 Chr.17: 41514020-41514020 on Build GRCh38
rs772936776 Chr.17: 41514123-41514123 on Build GRCh38
rs141578539 Chr.17: 41514170-41514170 on Build GRCh38
rs746030822 Chr.17: 41514054-41514054 on Build GRCh38
rs377111328 Chr.17: 41514038-41514038 on Build GRCh38
rs766239855 Chr.17: 41513997-41513997 on Build GRCh38
rs1013402951 Chr.17: 41514155-41514155 on Build GRCh38
rs1044433169 Chr.17: 41514163-41514163 on Build GRCh38
rs776693389 Chr.17: 41514078-41514078 on Build GRCh38
rs147870590 Chr.17: 41514164-41514164 on Build GRCh38
rs769138744 Chr.17: 41514065-41514065 on Build GRCh38
rs1031555890 Chr.17: 41514185-41514185 on Build GRCh38
rs754196440 Chr.17: 41514161-41514161 on Build GRCh38
rs779246507 Chr.17: 41514090-41514091 on Build GRCh38
rs780454273 Chr.17: 41514160-41514160 on Build GRCh38
rs761429493 Chr.17: 41513979-41513979 on Build GRCh38
rs969312306 Chr.17: 41514168-41514168 on Build GRCh38
rs536009613 Chr.17: 41514039-41514039 on Build GRCh38
rs372529305 Chr.17: 41514003-41514003 on Build GRCh38
rs752520098 Chr.17: 41514152-41514152 on Build GRCh38
rs753869748 Chr.17: 41514002-41514002 on Build GRCh38
rs757217904 Chr.17: 41514037-41514037 on Build GRCh38
rs368206269 Chr.17: 41514153-41514153 on Build GRCh38
rs895924375 Chr.17: 41514150-41514150 on Build GRCh38
rs143418415 Chr.17: 41514158-41514158 on Build GRCh38
rs910793725 Chr.17: 41514016-41514016 on Build GRCh38
rs745998616 Chr.17: 41514123-41514124 on Build GRCh38
rs371387812 Chr.17: 41513987-41513987 on Build GRCh38
rs943643386 Chr.17: 41514022-41514022 on Build GRCh38
rs891682713 Chr.17: 41514192-41514192 on Build GRCh38
rs560627933 Chr.17: 41514058-41514058 on Build GRCh38
rs756501099 Chr.17: 41514057-41514057 on Build GRCh38
rs770902579 Chr.17: 41514137-41514137 on Build GRCh38
rs764331835 Chr.17: 41514007-41514007 on Build GRCh38
rs759617949 Chr.17: 41514148-41514148 on Build GRCh38
rs552958702 Chr.17: 41514157-41514157 on Build GRCh38
rs760374799 Chr.17: 41514124-41514124 on Build GRCh38
rs765224814 Chr.17: 41514149-41514149 on Build GRCh38
rs772670883 Chr.17: 41513988-41513988 on Build GRCh38
rs1032104734 Chr.17: 41514180-41514180 on Build GRCh38
rs776558050 Chr.17: 41514144-41514144 on Build GRCh38
rs760529714 Chr.17: 41513994-41513994 on Build GRCh38
rs747695154 Chr.17: 41514096-41514096 on Build GRCh38

View Full Product Details