Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 946457-946684 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOC2L
Entrez Gene ID
26155
Gene Name
NOC2 like nucleolar associated transcriptional repressor
Gene Aliases
NET15, NET7, NIR, PPP1R112
UniGene
Hs.405987
Gene Chromosome Location
Chr.1: 944203-959299 on Build GRCh38
Transcripts
NM_015658.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7379936 c.1691C>T p.A564V Chr.1: 946514-946514 on Build GRCh38
4471133 c.1701C>T p.C567C Chr.1: 946504-946504 on Build GRCh38
7379934 c.1713G>T p.Q571H Chr.1: 946492-946492 on Build GRCh38
4434024 c.1710G>T p.V570V Chr.1: 946495-946495 on Build GRCh38
6024628 c.1716G>A p.Q572Q Chr.1: 946489-946489 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34516061 Chr.1: 946526-946527 on Build GRCh38
rs745356950 Chr.1: 946557-946558 on Build GRCh38
rs775671055 Chr.1: 946524-946524 on Build GRCh38
rs548320920 Chr.1: 946624-946624 on Build GRCh38
rs757860078 Chr.1: 946657-946657 on Build GRCh38
rs901788033 Chr.1: 946511-946511 on Build GRCh38
rs902223495 Chr.1: 946523-946523 on Build GRCh38
rs763357220 Chr.1: 946530-946530 on Build GRCh38
rs973746476 Chr.1: 946495-946495 on Build GRCh38
rs765132800 Chr.1: 946534-946534 on Build GRCh38
rs1057514740 Chr.1: 946596-946596 on Build GRCh38
rs531417864 Chr.1: 946618-946618 on Build GRCh38
rs943258910 Chr.1: 946482-946482 on Build GRCh38
rs749857767 Chr.1: 946505-946505 on Build GRCh38
rs1005062956 Chr.1: 946626-946626 on Build GRCh38
rs71628925 Chr.1: 946480-946480 on Build GRCh38
rs748596490 Chr.1: 946586-946586 on Build GRCh38
rs2272756 Chr.1: 946653-946653 on Build GRCh38
rs35471880 Chr.1: 946538-946538 on Build GRCh38
rs780604113 Chr.1: 946574-946574 on Build GRCh38
rs905889907 Chr.1: 946571-946571 on Build GRCh38
rs372813385 Chr.1: 946502-946502 on Build GRCh38
rs562211449 Chr.1: 946607-946607 on Build GRCh38
rs780563905 Chr.1: 946559-946559 on Build GRCh38
rs762004275 Chr.1: 946520-946520 on Build GRCh38
rs200220074 Chr.1: 946514-946514 on Build GRCh38
rs371122211 Chr.1: 946560-946560 on Build GRCh38
rs779250613 Chr.1: 946579-946579 on Build GRCh38
rs774955688 Chr.1: 946516-946516 on Build GRCh38
rs370843542 Chr.1: 946549-946549 on Build GRCh38
rs374468359 Chr.1: 946582-946582 on Build GRCh38
rs745447109 Chr.1: 946496-946496 on Build GRCh38
rs760797512 Chr.1: 946532-946532 on Build GRCh38
rs531963306 Chr.1: 946583-946583 on Build GRCh38
rs771983247 Chr.1: 946529-946529 on Build GRCh38
rs574093600 Chr.1: 946637-946637 on Build GRCh38
rs1053834077 Chr.1: 946531-946531 on Build GRCh38
rs755505231 Chr.1: 946558-946558 on Build GRCh38
rs867685413 Chr.1: 946506-946506 on Build GRCh38
rs200989028 Chr.1: 946550-946550 on Build GRCh38
rs886652308 Chr.1: 946603-946603 on Build GRCh38
rs776310067 Chr.1: 946621-946621 on Build GRCh38
rs752461609 Chr.1: 946537-946537 on Build GRCh38
rs756795890 Chr.1: 946554-946554 on Build GRCh38
rs780911631 Chr.1: 946492-946492 on Build GRCh38
rs138870888 Chr.1: 946584-946584 on Build GRCh38
rs755700468 Chr.1: 946561-946561 on Build GRCh38
rs909309836 Chr.1: 946625-946625 on Build GRCh38
rs201757857 Chr.1: 946503-946503 on Build GRCh38

View Full Product Details