Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 215945520-215945791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MREG
Entrez Gene ID
55686
Gene Name
melanoregulin
Gene Aliases
DSU, WDT2
UniGene
Hs.620391
Gene Chromosome Location
Chr.2: 215939097-216013623 on Build GRCh38
Transcripts
NM_018000.2 XM_011511468.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5808626 c.489A>G p.S163S Chr.2: 215945592-215945592 on Build GRCh38
4776733 c.494G>A p.R165K Chr.2: 215945587-215945587 on Build GRCh38
4091311 c.405T>G p.D135E Chr.2: 215945676-215945676 on Build GRCh38
4091310 c.427C>T p.R143* Chr.2: 215945654-215945654 on Build GRCh38
1719527 c.470C>A p.P157Q Chr.2: 215945611-215945611 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs942499129 Chr.2: 215945620-215945620 on Build GRCh38
rs779565613 Chr.2: 215945709-215945709 on Build GRCh38
rs762363894 Chr.2: 215945604-215945604 on Build GRCh38
rs375677015 Chr.2: 215945618-215945618 on Build GRCh38
rs766497457 Chr.2: 215945625-215945625 on Build GRCh38
rs757144666 Chr.2: 215945678-215945678 on Build GRCh38
rs993456075 Chr.2: 215945563-215945565 on Build GRCh38
rs909824562 Chr.2: 215945576-215945576 on Build GRCh38
rs374236068 Chr.2: 215945753-215945753 on Build GRCh38
rs764144154 Chr.2: 215945654-215945654 on Build GRCh38
rs775936253 Chr.2: 215945581-215945581 on Build GRCh38
rs868664178 Chr.2: 215945656-215945656 on Build GRCh38
rs984465189 Chr.2: 215945626-215945626 on Build GRCh38
rs34645939 Chr.2: 215945660-215945661 on Build GRCh38
rs971700321 Chr.2: 215945549-215945549 on Build GRCh38
rs554250823 Chr.2: 215945720-215945720 on Build GRCh38
rs200235388 Chr.2: 215945563-215945563 on Build GRCh38
rs368083579 Chr.2: 215945760-215945760 on Build GRCh38
rs779383794 Chr.2: 215945713-215945713 on Build GRCh38
rs749598732 Chr.2: 215945588-215945588 on Build GRCh38
rs756739984 Chr.2: 215945559-215945559 on Build GRCh38
rs62183184 Chr.2: 215945745-215945745 on Build GRCh38
rs923488831 Chr.2: 215945702-215945702 on Build GRCh38
rs747597078 Chr.2: 215945763-215945763 on Build GRCh38
rs780630202 Chr.2: 215945561-215945561 on Build GRCh38
rs765087884 Chr.2: 215945546-215945546 on Build GRCh38
rs572671930 Chr.2: 215945589-215945589 on Build GRCh38
rs751518395 Chr.2: 215945665-215945665 on Build GRCh38
rs371062715 Chr.2: 215945644-215945644 on Build GRCh38
rs749877230 Chr.2: 215945684-215945684 on Build GRCh38
rs773363112 Chr.2: 215945613-215945613 on Build GRCh38
rs370396010 Chr.2: 215945553-215945553 on Build GRCh38
rs148365522 Chr.2: 215945611-215945611 on Build GRCh38
rs375394259 Chr.2: 215945686-215945686 on Build GRCh38
rs759756929 Chr.2: 215945756-215945756 on Build GRCh38
rs368431426 Chr.2: 215945710-215945710 on Build GRCh38
rs762626563 Chr.2: 215945651-215945651 on Build GRCh38
rs780683542 Chr.2: 215945680-215945680 on Build GRCh38
rs772653014 Chr.2: 215945735-215945735 on Build GRCh38
rs774687123 Chr.2: 215945591-215945591 on Build GRCh38
rs763825310 Chr.2: 215945759-215945759 on Build GRCh38
rs372046464 Chr.2: 215945765-215945765 on Build GRCh38
rs746563861 Chr.2: 215945555-215945555 on Build GRCh38

View Full Product Details