Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2526709-2526958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LFNG
Entrez Gene ID
3955
Gene Name
LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Gene Aliases
SCDO3
UniGene
Hs.159142
Gene Chromosome Location
Chr.7: 2512529-2529177 on Build GRCh38
Transcripts
NM_001040167.1 NM_001040168.1 NM_001166355.1 NM_002304.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669658 c.1070C>T p.S357F Chr.7: 2526918-2526918 on Build GRCh38
4777006 c.1050C>G p.F350L Chr.7: 2526898-2526898 on Build GRCh38
4777008 c.663C>G p.F221L Chr.7: 2526898-2526898 on Build GRCh38
3669659 c.857C>T p.S286F Chr.7: 2526918-2526918 on Build GRCh38
4520083 c.828G>A p.K276K Chr.7: 2526889-2526889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769039241 Chr.7: 2526913-2526913 on Build GRCh38
rs756592084 Chr.7: 2526874-2526874 on Build GRCh38
rs770620949 Chr.7: 2526861-2526861 on Build GRCh38
rs541709346 Chr.7: 2526739-2526739 on Build GRCh38
rs780649312 Chr.7: 2526911-2526911 on Build GRCh38
rs201290357 Chr.7: 2526870-2526870 on Build GRCh38
rs751281684 Chr.7: 2526901-2526901 on Build GRCh38
rs376736772 Chr.7: 2526820-2526820 on Build GRCh38
rs1026508657 Chr.7: 2526778-2526778 on Build GRCh38
rs890205060 Chr.7: 2526818-2526818 on Build GRCh38
rs767620142 Chr.7: 2526830-2526830 on Build GRCh38
rs772085474 Chr.7: 2526883-2526883 on Build GRCh38
rs755459874 Chr.7: 2526935-2526935 on Build GRCh38
rs61743870 Chr.7: 2526850-2526850 on Build GRCh38
rs749905352 Chr.7: 2526910-2526910 on Build GRCh38
rs13225716 Chr.7: 2526799-2526799 on Build GRCh38
rs371194688 Chr.7: 2526810-2526810 on Build GRCh38
rs71647813 Chr.7: 2526884-2526884 on Build GRCh38
rs746375373 Chr.7: 2526813-2526813 on Build GRCh38
rs757790158 Chr.7: 2526809-2526809 on Build GRCh38
rs897490205 Chr.7: 2526747-2526747 on Build GRCh38
rs993534557 Chr.7: 2526784-2526784 on Build GRCh38
rs890499442 Chr.7: 2526733-2526733 on Build GRCh38
rs1001897517 Chr.7: 2526933-2526933 on Build GRCh38
rs868581959 Chr.7: 2526834-2526834 on Build GRCh38
rs200232463 Chr.7: 2526824-2526824 on Build GRCh38
rs752441376 Chr.7: 2526853-2526853 on Build GRCh38
rs199722773 Chr.7: 2526825-2526825 on Build GRCh38
rs147110861 Chr.7: 2526869-2526869 on Build GRCh38
rs763674563 Chr.7: 2526868-2526868 on Build GRCh38
rs141903867 Chr.7: 2526856-2526856 on Build GRCh38
rs764991226 Chr.7: 2526851-2526851 on Build GRCh38
rs763866787 Chr.7: 2526900-2526900 on Build GRCh38
rs762681069 Chr.7: 2526893-2526893 on Build GRCh38
rs775273178 Chr.7: 2526887-2526887 on Build GRCh38
rs139411864 Chr.7: 2526828-2526828 on Build GRCh38
rs761502412 Chr.7: 2526907-2526907 on Build GRCh38
rs746486395 Chr.7: 2526804-2526804 on Build GRCh38
rs766956135 Chr.7: 2526909-2526909 on Build GRCh38
rs749597262 Chr.7: 2526877-2526877 on Build GRCh38
rs777288540 Chr.7: 2526803-2526803 on Build GRCh38
rs372074206 Chr.7: 2526934-2526934 on Build GRCh38
rs138320196 Chr.7: 2526875-2526875 on Build GRCh38
rs143802425 Chr.7: 2526878-2526878 on Build GRCh38
rs886678544 Chr.7: 2526783-2526783 on Build GRCh38
rs775971890 Chr.7: 2526821-2526821 on Build GRCh38
rs771927828 Chr.7: 2526765-2526765 on Build GRCh38
rs201458148 Chr.7: 2526798-2526798 on Build GRCh38
rs370381985 Chr.7: 2526841-2526841 on Build GRCh38
rs748379564 Chr.7: 2526796-2526798 on Build GRCh38
rs1037998011 Chr.7: 2526746-2526746 on Build GRCh38
rs748395052 Chr.7: 2526880-2526880 on Build GRCh38

View Full Product Details