Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52848766-52848993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT78
Entrez Gene ID
196374
Gene Name
keratin 78
Gene Aliases
CK-78, K5B, K78, Kb40
UniGene
Hs.665267
Gene Chromosome Location
Chr.12: 52837804-52848994 on Build GRCh38
Transcripts
NM_173352.3 XM_011538010.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4899743 c.154C>T p.R52C Chr.12: 52848777-52848777 on Build GRCh38
4696891 c.22G>A p.A8T Chr.12: 52848909-52848909 on Build GRCh38
7198451 c.105C>A p.G35G Chr.12: 52848826-52848826 on Build GRCh38
4042946 c.44G>A p.R15H Chr.12: 52848887-52848887 on Build GRCh38
1932080 c.53G>T p.C18F Chr.12: 52848878-52848878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758418128 Chr.12: 52848816-52848816 on Build GRCh38
rs747556210 Chr.12: 52848909-52848909 on Build GRCh38
rs775106171 Chr.12: 52848946-52848946 on Build GRCh38
rs764048708 Chr.12: 52848872-52848872 on Build GRCh38
rs777207028 Chr.12: 52848817-52848817 on Build GRCh38
rs960776257 Chr.12: 52848852-52848852 on Build GRCh38
rs753032163 Chr.12: 52848813-52848814 on Build GRCh38
rs35831649 Chr.12: 52848860-52848860 on Build GRCh38
rs563484094 Chr.12: 52848855-52848855 on Build GRCh38
rs766956811 Chr.12: 52848962-52848962 on Build GRCh38
rs758928394 Chr.12: 52848905-52848905 on Build GRCh38
rs201230964 Chr.12: 52848828-52848828 on Build GRCh38
rs2253800 Chr.12: 52848901-52848901 on Build GRCh38
rs375338968 Chr.12: 52848956-52848956 on Build GRCh38
rs61754161 Chr.12: 52848811-52848811 on Build GRCh38
rs5798243 Chr.12: 52848845-52848845 on Build GRCh38
rs760071877 Chr.12: 52848792-52848793 on Build GRCh38
rs921981161 Chr.12: 52848969-52848969 on Build GRCh38
rs762827127 Chr.12: 52848950-52848950 on Build GRCh38
rs548010577 Chr.12: 52848836-52848836 on Build GRCh38
rs200256832 Chr.12: 52848814-52848831 on Build GRCh38
rs144892551 Chr.12: 52848856-52848856 on Build GRCh38
rs780748969 Chr.12: 52848888-52848888 on Build GRCh38
rs764925016 Chr.12: 52848869-52848869 on Build GRCh38
rs779084346 Chr.12: 52848897-52848897 on Build GRCh38
rs114041054 Chr.12: 52848911-52848911 on Build GRCh38
rs746018631 Chr.12: 52848924-52848924 on Build GRCh38
rs759865188 Chr.12: 52848867-52848867 on Build GRCh38
rs756433684 Chr.12: 52848829-52848830 on Build GRCh38
rs202030816 Chr.12: 52848887-52848887 on Build GRCh38
rs773442956 Chr.12: 52848789-52848789 on Build GRCh38
rs373647019 Chr.12: 52848912-52848912 on Build GRCh38
rs761796444 Chr.12: 52848961-52848961 on Build GRCh38
rs199880767 Chr.12: 52848829-52848829 on Build GRCh38
rs2253798 Chr.12: 52848795-52848795 on Build GRCh38
rs761192739 Chr.12: 52848920-52848920 on Build GRCh38
rs751653006 Chr.12: 52848793-52848793 on Build GRCh38
rs142981237 Chr.12: 52848870-52848870 on Build GRCh38
rs779392774 Chr.12: 52848854-52848854 on Build GRCh38
rs776958900 Chr.12: 52848861-52848861 on Build GRCh38
rs770059970 Chr.12: 52848926-52848926 on Build GRCh38
rs771180780 Chr.12: 52848858-52848858 on Build GRCh38
rs1042884614 Chr.12: 52848868-52848868 on Build GRCh38
rs755806128 Chr.12: 52848892-52848892 on Build GRCh38
rs184419711 Chr.12: 52848798-52848798 on Build GRCh38
rs778137492 Chr.12: 52848830-52848847 on Build GRCh38
rs768898661 Chr.12: 52848851-52848851 on Build GRCh38
rs778405023 Chr.12: 52848906-52848906 on Build GRCh38
rs369107504 Chr.12: 52848891-52848891 on Build GRCh38
rs766741699 Chr.12: 52848797-52848797 on Build GRCh38
rs752569167 Chr.12: 52848814-52848814 on Build GRCh38
rs745576224 Chr.12: 52848846-52848846 on Build GRCh38
rs750684727 Chr.12: 52848949-52848949 on Build GRCh38
rs11170289 Chr.12: 52848857-52848857 on Build GRCh38
rs151107252 Chr.12: 52848875-52848875 on Build GRCh38
rs774269693 Chr.12: 52848959-52848959 on Build GRCh38
rs780801966 Chr.12: 52848842-52848842 on Build GRCh38

View Full Product Details