Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119834692-119834963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NBPF7
Entrez Gene ID
343505
Gene Name
neuroblastoma breakpoint family member 7
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 119833730-119844880 on Build GRCh38
Transcripts
NM_001047980.2 XM_017001186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6120840 c.1229C>T p.P410L Chr.1: 119834802-119834802 on Build GRCh38
334177 c.1188G>A p.L396L Chr.1: 119834843-119834843 on Build GRCh38
5605907 c.1146G>A p.K382K Chr.1: 119834885-119834885 on Build GRCh38
6817329 c.1202delA p.N401fs*>21 Chr.1: 119834829-119834829 on Build GRCh38
3417958 c.1224A>G p.S408S Chr.1: 119834807-119834807 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763350579 Chr.1: 119834911-119834911 on Build GRCh38
rs1056624770 Chr.1: 119834834-119834834 on Build GRCh38
rs778834173 Chr.1: 119834850-119834850 on Build GRCh38
rs766890320 Chr.1: 119834933-119834933 on Build GRCh38
rs750872342 Chr.1: 119834811-119834811 on Build GRCh38
rs770876021 Chr.1: 119834774-119834774 on Build GRCh38
rs772190738 Chr.1: 119834783-119834783 on Build GRCh38
rs375777967 Chr.1: 119834887-119834888 on Build GRCh38
rs765591751 Chr.1: 119834808-119834808 on Build GRCh38
rs587640368 Chr.1: 119834793-119834793 on Build GRCh38
rs770059414 Chr.1: 119834901-119834901 on Build GRCh38
rs1009835529 Chr.1: 119834843-119834843 on Build GRCh38
rs760707351 Chr.1: 119834792-119834792 on Build GRCh38
rs776740768 Chr.1: 119834794-119834794 on Build GRCh38
rs201802573 Chr.1: 119834797-119834797 on Build GRCh38
rs201245200 Chr.1: 119834764-119834764 on Build GRCh38
rs774395335 Chr.1: 119834777-119834777 on Build GRCh38
rs775537396 Chr.1: 119834762-119834762 on Build GRCh38
rs747060604 Chr.1: 119834867-119834867 on Build GRCh38
rs587686506 Chr.1: 119834889-119834889 on Build GRCh38
rs775633499 Chr.1: 119834785-119834785 on Build GRCh38
rs587724206 Chr.1: 119834868-119834869 on Build GRCh38
rs766864452 Chr.1: 119834826-119834826 on Build GRCh38
rs777437159 Chr.1: 119834837-119834837 on Build GRCh38
rs762035595 Chr.1: 119834891-119834891 on Build GRCh38
rs879098510 Chr.1: 119834752-119834752 on Build GRCh38
rs905364771 Chr.1: 119834802-119834802 on Build GRCh38
rs768649123 Chr.1: 119834881-119834881 on Build GRCh38
rs908212926 Chr.1: 119834769-119834769 on Build GRCh38
rs376376773 Chr.1: 119834743-119834743 on Build GRCh38
rs780768485 Chr.1: 119834721-119834721 on Build GRCh38
rs762759352 Chr.1: 119834937-119834937 on Build GRCh38
rs777334253 Chr.1: 119834748-119834748 on Build GRCh38
rs779903465 Chr.1: 119834866-119834866 on Build GRCh38
rs758894052 Chr.1: 119834821-119834821 on Build GRCh38
rs200252233 Chr.1: 119834840-119834840 on Build GRCh38
rs963880516 Chr.1: 119834723-119834723 on Build GRCh38
rs991043760 Chr.1: 119834735-119834735 on Build GRCh38
rs587627604 Chr.1: 119834835-119834835 on Build GRCh38
rs587729856 Chr.1: 119834833-119834833 on Build GRCh38
rs374554492 Chr.1: 119834939-119834939 on Build GRCh38
rs370502677 Chr.1: 119834930-119834930 on Build GRCh38
rs933650929 Chr.1: 119834757-119834757 on Build GRCh38
rs374232683 Chr.1: 119834839-119834839 on Build GRCh38
rs772100823 Chr.1: 119834864-119834864 on Build GRCh38
rs746965744 Chr.1: 119834795-119834795 on Build GRCh38
rs745721644 Chr.1: 119834851-119834851 on Build GRCh38
rs922548545 Chr.1: 119834745-119834745 on Build GRCh38
rs776858385 Chr.1: 119834935-119834935 on Build GRCh38
rs371095005 Chr.1: 119834807-119834807 on Build GRCh38
rs758327526 Chr.1: 119834744-119834744 on Build GRCh38
rs1017735763 Chr.1: 119834905-119834905 on Build GRCh38
rs773563091 Chr.1: 119834904-119834904 on Build GRCh38
rs753453030 Chr.1: 119834941-119834941 on Build GRCh38
rs780708330 Chr.1: 119834855-119834855 on Build GRCh38
rs745809548 Chr.1: 119834779-119834779 on Build GRCh38

View Full Product Details