Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 151679714-151679987 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GATB
Entrez Gene ID
5188
Gene Name
glutamyl-tRNA amidotransferase subunit B
Gene Aliases
HSPC199, PET112, PET112L
UniGene
Hs.119316
Gene Chromosome Location
Chr.4: 151669773-151761023 on Build GRCh38
Transcripts
NM_004564.2 XM_005263063.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5865877 c.1334C>T p.P445L Chr.4: 151679889-151679889 on Build GRCh38
267165 c.1384A>C p.T462P Chr.4: 151679839-151679839 on Build GRCh38
5400627 c.1385C>T p.T462I Chr.4: 151679838-151679838 on Build GRCh38
6744047 c.1348G>A p.A450T Chr.4: 151679875-151679875 on Build GRCh38
1753568 c.1402G>A p.A468T Chr.4: 151679821-151679821 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751588690 Chr.4: 151679960-151679960 on Build GRCh38
rs747400337 Chr.4: 151679784-151679784 on Build GRCh38
rs769699220 Chr.4: 151679789-151679789 on Build GRCh38
rs974370064 Chr.4: 151679961-151679961 on Build GRCh38
rs772871311 Chr.4: 151679818-151679818 on Build GRCh38
rs751065403 Chr.4: 151679912-151679912 on Build GRCh38
rs764073602 Chr.4: 151679869-151679869 on Build GRCh38
rs553618128 Chr.4: 151679804-151679804 on Build GRCh38
rs770387773 Chr.4: 151679827-151679827 on Build GRCh38
rs754493564 Chr.4: 151679925-151679925 on Build GRCh38
rs779748513 Chr.4: 151679899-151679899 on Build GRCh38
rs759596188 Chr.4: 151679852-151679852 on Build GRCh38
rs1045690695 Chr.4: 151679820-151679820 on Build GRCh38
rs762998814 Chr.4: 151679959-151679959 on Build GRCh38
rs781355798 Chr.4: 151679803-151679803 on Build GRCh38
rs763286404 Chr.4: 151679834-151679834 on Build GRCh38
rs1008452941 Chr.4: 151679945-151679945 on Build GRCh38
rs1018470540 Chr.4: 151679952-151679952 on Build GRCh38
rs779668805 Chr.4: 151679777-151679777 on Build GRCh38
rs769071663 Chr.4: 151679796-151679796 on Build GRCh38
rs994668480 Chr.4: 151679900-151679900 on Build GRCh38
rs750020229 Chr.4: 151679888-151679888 on Build GRCh38
rs758095140 Chr.4: 151679773-151679773 on Build GRCh38
rs368156732 Chr.4: 151679940-151679940 on Build GRCh38
rs758050203 Chr.4: 151679891-151679891 on Build GRCh38
rs774488466 Chr.4: 151679843-151679843 on Build GRCh38
rs141826728 Chr.4: 151679849-151679849 on Build GRCh38
rs773469580 Chr.4: 151679830-151679830 on Build GRCh38
rs139428309 Chr.4: 151679836-151679836 on Build GRCh38
rs755499526 Chr.4: 151679857-151679858 on Build GRCh38
rs953824884 Chr.4: 151679898-151679898 on Build GRCh38
rs761631015 Chr.4: 151679870-151679870 on Build GRCh38
rs748713795 Chr.4: 151679823-151679823 on Build GRCh38
rs375643617 Chr.4: 151679872-151679872 on Build GRCh38
rs572270367 Chr.4: 151679769-151679769 on Build GRCh38
rs371087303 Chr.4: 151679770-151679770 on Build GRCh38

View Full Product Details