Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2586135-2586396 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCE
Entrez Gene ID
23288
Gene Name
IQ motif containing E
Gene Aliases
1700028P05Rik
UniGene
Hs.520627
Gene Chromosome Location
Chr.7: 2558972-2614734 on Build GRCh38
Transcripts
NM_001287499.1 NM_001287500.1 NM_001287501.1 NM_001287502.1 NM_152558.4 XM_006715676.2 XM_011515242.1 XM_011515243.1 XM_011515244.2 XM_017011902.1 XM_017011903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038144 c.825-4C>T p.? Chr.7: 2586204-2586204 on Build GRCh38
3880363 c.850G>A p.A284T Chr.7: 2586233-2586233 on Build GRCh38
1088810 c.951C>T p.R317R Chr.7: 2586334-2586334 on Build GRCh38
6261354 c.929G>T p.S310I Chr.7: 2586312-2586312 on Build GRCh38
3950364 c.893C>T p.S298F Chr.7: 2586276-2586276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775981076 Chr.7: 2586225-2586225 on Build GRCh38
rs764921146 Chr.7: 2586369-2586398 on Build GRCh38
rs560190603 Chr.7: 2586299-2586299 on Build GRCh38
rs758125778 Chr.7: 2586258-2586258 on Build GRCh38
rs771444829 Chr.7: 2586167-2586167 on Build GRCh38
rs777080126 Chr.7: 2586170-2586170 on Build GRCh38
rs201196475 Chr.7: 2586228-2586228 on Build GRCh38
rs376928054 Chr.7: 2586211-2586211 on Build GRCh38
rs770376079 Chr.7: 2586209-2586209 on Build GRCh38
rs925435325 Chr.7: 2586284-2586284 on Build GRCh38
rs780744463 Chr.7: 2586292-2586292 on Build GRCh38
rs770196675 Chr.7: 2586218-2586218 on Build GRCh38
rs774115233 Chr.7: 2586255-2586255 on Build GRCh38
rs745821447 Chr.7: 2586197-2586197 on Build GRCh38
rs368939000 Chr.7: 2586355-2586355 on Build GRCh38
rs547114342 Chr.7: 2586278-2586278 on Build GRCh38
rs200208942 Chr.7: 2586368-2586368 on Build GRCh38
rs267601471 Chr.7: 2586265-2586265 on Build GRCh38
rs557825712 Chr.7: 2586369-2586369 on Build GRCh38
rs374856058 Chr.7: 2586295-2586295 on Build GRCh38
rs191503365 Chr.7: 2586214-2586214 on Build GRCh38
rs780931746 Chr.7: 2586195-2586195 on Build GRCh38
rs2257285 Chr.7: 2586204-2586204 on Build GRCh38
rs373779920 Chr.7: 2586206-2586206 on Build GRCh38
rs199981463 Chr.7: 2586232-2586232 on Build GRCh38
rs147147454 Chr.7: 2586191-2586191 on Build GRCh38
rs765076819 Chr.7: 2586237-2586237 on Build GRCh38
rs750439401 Chr.7: 2586233-2586233 on Build GRCh38
rs763167432 Chr.7: 2586227-2586227 on Build GRCh38
rs186271009 Chr.7: 2586196-2586196 on Build GRCh38
rs762167587 Chr.7: 2586322-2586322 on Build GRCh38
rs765678803 Chr.7: 2586175-2586175 on Build GRCh38
rs78104931 Chr.7: 2586332-2586332 on Build GRCh38
rs749854999 Chr.7: 2586298-2586298 on Build GRCh38
rs769095246 Chr.7: 2586318-2586318 on Build GRCh38
rs780649606 Chr.7: 2586371-2586371 on Build GRCh38
rs756868544 Chr.7: 2586279-2586279 on Build GRCh38
rs760167231 Chr.7: 2586157-2586157 on Build GRCh38
rs372247306 Chr.7: 2586372-2586372 on Build GRCh38
rs749693208 Chr.7: 2586309-2586309 on Build GRCh38
rs765228487 Chr.7: 2586334-2586334 on Build GRCh38
rs773701437 Chr.7: 2586273-2586282 on Build GRCh38
rs756960597 Chr.7: 2586360-2586360 on Build GRCh38
rs61736928 Chr.7: 2586262-2586262 on Build GRCh38
rs549595786 Chr.7: 2586344-2586344 on Build GRCh38
rs201181062 Chr.7: 2586319-2586319 on Build GRCh38
rs1013594173 Chr.7: 2586241-2586241 on Build GRCh38
rs755012927 Chr.7: 2586235-2586235 on Build GRCh38
rs569845935 Chr.7: 2586350-2586350 on Build GRCh38
rs752969846 Chr.7: 2586182-2586182 on Build GRCh38
rs761022107 Chr.7: 2586338-2586338 on Build GRCh38
rs767849059 Chr.7: 2586331-2586331 on Build GRCh38
rs751792535 Chr.7: 2586229-2586229 on Build GRCh38
rs771819944 Chr.7: 2586200-2586200 on Build GRCh38
rs755210717 Chr.7: 2586174-2586174 on Build GRCh38
rs755539386 Chr.7: 2586375-2586375 on Build GRCh38
rs78092993 Chr.7: 2586185-2586185 on Build GRCh38
rs770348134 Chr.7: 2586297-2586297 on Build GRCh38
rs180735566 Chr.7: 2586333-2586333 on Build GRCh38

View Full Product Details