Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 133753724-133753989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPC3
Entrez Gene ID
2719
Gene Name
glypican 3
Gene Aliases
DGSX, GTR2-2, MXR7, OCI-5, SDYS, SGB, SGBS, SGBS1
UniGene
Hs.644108
Gene Chromosome Location
Chr.X: 133535745-133985646 on Build GRCh38
Transcripts
NM_001164617.1 NM_001164618.1 NM_001164619.1 NM_004484.3 XM_017029413.1

SNP Detail

SNP ID Chromosome Location
rs774682860 Chr.X: 133753832-133753832 on Build GRCh38
rs900143355 Chr.X: 133753949-133753949 on Build GRCh38
rs756480292 Chr.X: 133753907-133753907 on Build GRCh38
rs773264161 Chr.X: 133753841-133753841 on Build GRCh38
rs746527435 Chr.X: 133753751-133753751 on Build GRCh38
rs756693876 Chr.X: 133753934-133753934 on Build GRCh38
rs201344338 Chr.X: 133753912-133753912 on Build GRCh38
rs745317547 Chr.X: 133753930-133753930 on Build GRCh38
rs866197025 Chr.X: 133753811-133753811 on Build GRCh38
rs140848049 Chr.X: 133753890-133753890 on Build GRCh38
rs776010902 Chr.X: 133753779-133753779 on Build GRCh38
rs780667597 Chr.X: 133753945-133753945 on Build GRCh38
rs199778216 Chr.X: 133753781-133753781 on Build GRCh38
rs752194694 Chr.X: 133753754-133753754 on Build GRCh38
rs767701856 Chr.X: 133753840-133753840 on Build GRCh38
rs182960589 Chr.X: 133753746-133753746 on Build GRCh38
rs373406834 Chr.X: 133753842-133753842 on Build GRCh38
rs768782219 Chr.X: 133753821-133753821 on Build GRCh38
rs763779764 Chr.X: 133753899-133753899 on Build GRCh38
rs752516966 Chr.X: 133753866-133753866 on Build GRCh38
rs145141525 Chr.X: 133753838-133753838 on Build GRCh38
rs780908710 Chr.X: 133753913-133753913 on Build GRCh38
rs749735623 Chr.X: 133753961-133753961 on Build GRCh38
rs112530065 Chr.X: 133753843-133753843 on Build GRCh38
rs138450923 Chr.X: 133753854-133753854 on Build GRCh38
rs770414070 Chr.X: 133753761-133753761 on Build GRCh38
rs375160737 Chr.X: 133753906-133753906 on Build GRCh38
rs751267769 Chr.X: 133753904-133753904 on Build GRCh38
rs104894855 Chr.X: 133753919-133753919 on Build GRCh38
rs934493521 Chr.X: 133753752-133753752 on Build GRCh38
rs756844729 Chr.X: 133753753-133753753 on Build GRCh38

View Full Product Details