Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 161042482-161042753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USF1
Entrez Gene ID
7391
Gene Name
upstream transcription factor 1
Gene Aliases
FCHL, FCHL1, HYPLIP1, MLTF, MLTFI, UEF, bHLHb11
UniGene
Hs.414880
Gene Chromosome Location
Chr.1: 161039251-161045979 on Build GRCh38
Transcripts
NM_001276373.1 NM_007122.4 NM_207005.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5844228 c.147C>A p.Y49* Chr.1: 161042582-161042582 on Build GRCh38
6995393 c.125delT p.F42fs*18 Chr.1: 161042604-161042604 on Build GRCh38
2151237 c.71C>A p.T24N Chr.1: 161042658-161042658 on Build GRCh38
98056 c.148G>A p.V50I Chr.1: 161042581-161042581 on Build GRCh38
6265505 c.105C>A p.S35R Chr.1: 161042624-161042624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773103510 Chr.1: 161042650-161042650 on Build GRCh38
rs753237731 Chr.1: 161042715-161042715 on Build GRCh38
rs142286568 Chr.1: 161042613-161042613 on Build GRCh38
rs755706110 Chr.1: 161042596-161042596 on Build GRCh38
rs775218430 Chr.1: 161042536-161042552 on Build GRCh38
rs763506914 Chr.1: 161042602-161042602 on Build GRCh38
rs756800992 Chr.1: 161042605-161042605 on Build GRCh38
rs34882696 Chr.1: 161042703-161042703 on Build GRCh38
rs748592178 Chr.1: 161042639-161042639 on Build GRCh38
rs200939414 Chr.1: 161042562-161042562 on Build GRCh38
rs376077931 Chr.1: 161042723-161042723 on Build GRCh38
rs756358871 Chr.1: 161042538-161042538 on Build GRCh38
rs765022880 Chr.1: 161042720-161042720 on Build GRCh38
rs913737325 Chr.1: 161042637-161042637 on Build GRCh38
rs532379635 Chr.1: 161042517-161042517 on Build GRCh38
rs17175505 Chr.1: 161042724-161042724 on Build GRCh38
rs199784242 Chr.1: 161042658-161042658 on Build GRCh38
rs896457394 Chr.1: 161042734-161042734 on Build GRCh38
rs199606185 Chr.1: 161042642-161042642 on Build GRCh38
rs905149427 Chr.1: 161042694-161042694 on Build GRCh38
rs767480001 Chr.1: 161042591-161042591 on Build GRCh38
rs1026130854 Chr.1: 161042558-161042558 on Build GRCh38
rs759372316 Chr.1: 161042671-161042671 on Build GRCh38
rs568694042 Chr.1: 161042719-161042719 on Build GRCh38
rs780498058 Chr.1: 161042513-161042513 on Build GRCh38
rs933756790 Chr.1: 161042626-161042626 on Build GRCh38
rs747337778 Chr.1: 161042614-161042614 on Build GRCh38
rs375213662 Chr.1: 161042606-161042606 on Build GRCh38
rs752606775 Chr.1: 161042592-161042592 on Build GRCh38
rs761995109 Chr.1: 161042521-161042521 on Build GRCh38
rs755365665 Chr.1: 161042629-161042629 on Build GRCh38
rs771402225 Chr.1: 161042522-161042522 on Build GRCh38
rs763085474 Chr.1: 161042543-161042543 on Build GRCh38
rs985059911 Chr.1: 161042527-161042527 on Build GRCh38
rs767284073 Chr.1: 161042673-161042673 on Build GRCh38
rs753532389 Chr.1: 161042603-161042603 on Build GRCh38
rs369938502 Chr.1: 161042709-161042709 on Build GRCh38
rs749543020 Chr.1: 161042652-161042652 on Build GRCh38
rs1043558002 Chr.1: 161042653-161042653 on Build GRCh38
rs759496934 Chr.1: 161042582-161042582 on Build GRCh38
rs564660292 Chr.1: 161042697-161042697 on Build GRCh38
rs776883166 Chr.1: 161042520-161042520 on Build GRCh38
rs774522606 Chr.1: 161042670-161042670 on Build GRCh38
rs372461061 Chr.1: 161042633-161042633 on Build GRCh38
rs552690907 Chr.1: 161042550-161042550 on Build GRCh38
rs769742062 Chr.1: 161042528-161042528 on Build GRCh38
rs1056506127 Chr.1: 161042732-161042732 on Build GRCh38
rs878952764 Chr.1: 161042549-161042549 on Build GRCh38
rs964931505 Chr.1: 161042575-161042575 on Build GRCh38

View Full Product Details