Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 15189725-15189956 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39B
Entrez Gene ID
158219
Gene Name
tetratricopeptide repeat domain 39B
Gene Aliases
C9orf52
UniGene
Hs.563630
Gene Chromosome Location
Chr.9: 15167204-15307360 on Build GRCh38
Transcripts
NM_001168339.1 NM_001168340.1 NM_001168341.1 NM_001168342.1 NM_152574.2 XM_011517732.2 XM_017014310.1 XM_017014311.1 XM_017014312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7297730 c.1138C>T p.R380C Chr.9: 15189760-15189760 on Build GRCh38
1330622 c.929C>A p.A310E Chr.9: 15189771-15189771 on Build GRCh38
4372907 c.1107T>C p.G369G Chr.9: 15189791-15189791 on Build GRCh38
3090462 c.1107T>C p.G369G Chr.9: 15189791-15189791 on Build GRCh38
1330621 c.1127C>A p.A376E Chr.9: 15189771-15189771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772450392 Chr.9: 15189800-15189800 on Build GRCh38
rs61755267 Chr.9: 15189759-15189759 on Build GRCh38
rs558759098 Chr.9: 15189837-15189837 on Build GRCh38
rs959222912 Chr.9: 15189764-15189764 on Build GRCh38
rs757749130 Chr.9: 15189874-15189874 on Build GRCh38
rs752634264 Chr.9: 15189768-15189768 on Build GRCh38
rs35550341 Chr.9: 15189829-15189830 on Build GRCh38
rs968363706 Chr.9: 15189785-15189785 on Build GRCh38
rs776950748 Chr.9: 15189821-15189821 on Build GRCh38
rs536859209 Chr.9: 15189833-15189833 on Build GRCh38
rs376726304 Chr.9: 15189770-15189770 on Build GRCh38
rs143112996 Chr.9: 15189771-15189771 on Build GRCh38
rs975831502 Chr.9: 15189820-15189820 on Build GRCh38
rs748548687 Chr.9: 15189797-15189797 on Build GRCh38
rs766497263 Chr.9: 15189754-15189754 on Build GRCh38
rs754328219 Chr.9: 15189760-15189760 on Build GRCh38
rs774673803 Chr.9: 15189794-15189794 on Build GRCh38
rs766483713 Chr.9: 15189819-15189819 on Build GRCh38
rs756853041 Chr.9: 15189775-15189775 on Build GRCh38
rs765607152 Chr.9: 15189827-15189827 on Build GRCh38
rs907287702 Chr.9: 15189750-15189750 on Build GRCh38
rs932205921 Chr.9: 15189883-15189883 on Build GRCh38
rs745526289 Chr.9: 15189788-15189788 on Build GRCh38
rs768115369 Chr.9: 15189747-15189747 on Build GRCh38
rs576049921 Chr.9: 15189777-15189777 on Build GRCh38
rs750435177 Chr.9: 15189749-15189749 on Build GRCh38
rs868160948 Chr.9: 15189884-15189884 on Build GRCh38
rs376568277 Chr.9: 15189832-15189832 on Build GRCh38
rs940900014 Chr.9: 15189923-15189923 on Build GRCh38
rs541369274 Chr.9: 15189917-15189917 on Build GRCh38
rs371588203 Chr.9: 15189825-15189825 on Build GRCh38
rs577224003 Chr.9: 15189844-15189844 on Build GRCh38
rs773826461 Chr.9: 15189805-15189805 on Build GRCh38
rs1002934612 Chr.9: 15189758-15189758 on Build GRCh38
rs760871760 Chr.9: 15189751-15189751 on Build GRCh38
rs768957181 Chr.9: 15189792-15189792 on Build GRCh38
rs765153413 Chr.9: 15189763-15189763 on Build GRCh38
rs760612555 Chr.9: 15189810-15189810 on Build GRCh38
rs369233965 Chr.9: 15189757-15189757 on Build GRCh38

View Full Product Details