Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133101927-133102170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RALGDS
Entrez Gene ID
5900
Gene Name
ral guanine nucleotide dissociation stimulator
Gene Aliases
RGDS, RGF, RalGEF
UniGene
Hs.106185
Gene Chromosome Location
Chr.9: 133097720-133149220 on Build GRCh38
Transcripts
NM_001042368.2 NM_001271774.1 NM_001271775.1 NM_001271776.1 NM_006266.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6027709 c.2050G>A p.G684S Chr.9: 133102012-133102012 on Build GRCh38
7331119 c.2421G>C p.K807N Chr.9: 133101941-133101941 on Build GRCh38
4639622 c.1980T>C p.C660C Chr.9: 133102082-133102082 on Build GRCh38
6850485 c.2186A>C p.E729A Chr.9: 133101963-133101963 on Build GRCh38
6322975 c.2109C>A p.I703I Chr.9: 133102040-133102040 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053204114 Chr.9: 133102110-133102110 on Build GRCh38
rs139082000 Chr.9: 133102092-133102092 on Build GRCh38
rs373357690 Chr.9: 133102017-133102017 on Build GRCh38
rs764119929 Chr.9: 133102024-133102024 on Build GRCh38
rs376306097 Chr.9: 133102048-133102048 on Build GRCh38
rs34170541 Chr.9: 133101979-133101979 on Build GRCh38
rs774681232 Chr.9: 133101994-133101994 on Build GRCh38
rs767544998 Chr.9: 133101997-133101997 on Build GRCh38
rs113532379 Chr.9: 133102012-133102012 on Build GRCh38
rs35542708 Chr.9: 133102092-133102093 on Build GRCh38
rs537897228 Chr.9: 133102016-133102016 on Build GRCh38
rs772631258 Chr.9: 133102115-133102115 on Build GRCh38
rs916517753 Chr.9: 133102045-133102045 on Build GRCh38
rs35267131 Chr.9: 133102124-133102124 on Build GRCh38
rs925942592 Chr.9: 133102085-133102085 on Build GRCh38
rs748645363 Chr.9: 133102106-133102106 on Build GRCh38
rs141468667 Chr.9: 133102025-133102025 on Build GRCh38
rs1043106953 Chr.9: 133102068-133102068 on Build GRCh38
rs370960854 Chr.9: 133102032-133102032 on Build GRCh38
rs769716366 Chr.9: 133101986-133101988 on Build GRCh38
rs755889399 Chr.9: 133102033-133102033 on Build GRCh38
rs147006349 Chr.9: 133102039-133102039 on Build GRCh38
rs200199802 Chr.9: 133102040-133102040 on Build GRCh38
rs757194753 Chr.9: 133102030-133102030 on Build GRCh38
rs760693070 Chr.9: 133102119-133102119 on Build GRCh38
rs780888252 Chr.9: 133102031-133102031 on Build GRCh38
rs906554175 Chr.9: 133102088-133102088 on Build GRCh38
rs745562514 Chr.9: 133101953-133101953 on Build GRCh38
rs758059159 Chr.9: 133102020-133102020 on Build GRCh38
rs148398301 Chr.9: 133101966-133101966 on Build GRCh38
rs35831936 Chr.9: 133102013-133102013 on Build GRCh38
rs773247714 Chr.9: 133101998-133101998 on Build GRCh38
rs984770014 Chr.9: 133101983-133101983 on Build GRCh38
rs368487328 Chr.9: 133102123-133102123 on Build GRCh38
rs140751236 Chr.9: 133101995-133101995 on Build GRCh38
rs763841878 Chr.9: 133102127-133102127 on Build GRCh38
rs769538916 Chr.9: 133101965-133101965 on Build GRCh38
rs890507267 Chr.9: 133102117-133102117 on Build GRCh38
rs748411421 Chr.9: 133102137-133102137 on Build GRCh38
rs766588898 Chr.9: 133102122-133102122 on Build GRCh38
rs900782953 Chr.9: 133102138-133102138 on Build GRCh38
rs939206593 Chr.9: 133102034-133102034 on Build GRCh38
rs773342144 Chr.9: 133102118-133102118 on Build GRCh38
rs548531503 Chr.9: 133101970-133101970 on Build GRCh38
rs142584543 Chr.9: 133101977-133101977 on Build GRCh38
rs181672343 Chr.9: 133101948-133101948 on Build GRCh38
rs757207457 Chr.9: 133102148-133102148 on Build GRCh38
rs890621721 Chr.9: 133101989-133101989 on Build GRCh38

View Full Product Details