Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19588582-19588855 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3KBP1
Entrez Gene ID
30011
Gene Name
SH3 domain containing kinase binding protein 1
Gene Aliases
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
UniGene
Hs.726365
Gene Chromosome Location
Chr.X: 19533965-19887626 on Build GRCh38
Transcripts
NM_001024666.2 NM_001184960.1 NM_031892.2 XM_005274494.2 XM_011545498.2 XM_011545499.2 XM_011545500.2 XM_011545502.2 XM_017029459.1 XM_017029460.1 XM_017029461.1 XM_017029462.1 XM_017029463.1 XM_017029464.1 XM_017029465.1 XM_017029466.1 XM_017029467.1 XM_017029468.1 XM_017029469.1 XM_017029470.1

SNP Detail

SNP ID Chromosome Location
rs936840141 Chr.X: 19588728-19588728 on Build GRCh38
rs748530625 Chr.X: 19588756-19588756 on Build GRCh38
rs149782443 Chr.X: 19588758-19588758 on Build GRCh38
rs369373145 Chr.X: 19588621-19588621 on Build GRCh38
rs757441642 Chr.X: 19588615-19588615 on Build GRCh38
rs780311990 Chr.X: 19588741-19588741 on Build GRCh38
rs201278502 Chr.X: 19588704-19588704 on Build GRCh38
rs774272802 Chr.X: 19588731-19588731 on Build GRCh38
rs759595410 Chr.X: 19588744-19588744 on Build GRCh38
rs752483118 Chr.X: 19588685-19588685 on Build GRCh38
rs757468529 Chr.X: 19588686-19588686 on Build GRCh38
rs147648876 Chr.X: 19588666-19588666 on Build GRCh38
rs145308311 Chr.X: 19588657-19588657 on Build GRCh38
rs145759622 Chr.X: 19588759-19588759 on Build GRCh38
rs2290805 Chr.X: 19588675-19588675 on Build GRCh38
rs767293120 Chr.X: 19588676-19588676 on Build GRCh38
rs199796248 Chr.X: 19588684-19588684 on Build GRCh38
rs772695801 Chr.X: 19588620-19588620 on Build GRCh38
rs181448350 Chr.X: 19588625-19588625 on Build GRCh38
rs483352756 Chr.X: 19588683-19588683 on Build GRCh38
rs776886589 Chr.X: 19588648-19588648 on Build GRCh38
rs967435715 Chr.X: 19588811-19588811 on Build GRCh38
rs765884745 Chr.X: 19588667-19588667 on Build GRCh38
rs751426798 Chr.X: 19588672-19588672 on Build GRCh38
rs374793970 Chr.X: 19588650-19588650 on Build GRCh38
rs377076113 Chr.X: 19588616-19588616 on Build GRCh38
rs750689524 Chr.X: 19588717-19588717 on Build GRCh38
rs747523659 Chr.X: 19588622-19588622 on Build GRCh38
rs745983365 Chr.X: 19588617-19588617 on Build GRCh38
rs372490590 Chr.X: 19588604-19588604 on Build GRCh38
rs773873603 Chr.X: 19588807-19588807 on Build GRCh38
rs769375018 Chr.X: 19588658-19588658 on Build GRCh38
rs770384119 Chr.X: 19588789-19588789 on Build GRCh38
rs960905963 Chr.X: 19588689-19588689 on Build GRCh38

View Full Product Details