Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192576310-192576569 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS1
Entrez Gene ID
5996
Gene Name
regulator of G-protein signaling 1
Gene Aliases
1R20, BL34, HEL-S-87, IER1, IR20
UniGene
Hs.75256
Gene Chromosome Location
Chr.1: 192575727-192580029 on Build GRCh38
Transcripts
NM_002922.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3863924 c.195G>A p.M65I Chr.1: 192576342-192576342 on Build GRCh38
3863927 c.204C>T p.S68S Chr.1: 192576351-192576351 on Build GRCh38
6060270 c.145G>T p.E49* Chr.1: 192576331-192576331 on Build GRCh38
4471924 c.174C>T p.I58I Chr.1: 192576321-192576321 on Build GRCh38
3863928 c.165C>T p.S55S Chr.1: 192576351-192576351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs944183655 Chr.1: 192576426-192576426 on Build GRCh38
rs559762390 Chr.1: 192576431-192576431 on Build GRCh38
rs758026916 Chr.1: 192576347-192576347 on Build GRCh38
rs757618719 Chr.1: 192576373-192576373 on Build GRCh38
rs770844779 Chr.1: 192576449-192576449 on Build GRCh38
rs768973607 Chr.1: 192576391-192576391 on Build GRCh38
rs760613828 Chr.1: 192576409-192576409 on Build GRCh38
rs774683897 Chr.1: 192576406-192576406 on Build GRCh38
rs942652277 Chr.1: 192576365-192576365 on Build GRCh38
rs774498869 Chr.1: 192576455-192576455 on Build GRCh38
rs780939760 Chr.1: 192576368-192576368 on Build GRCh38
rs375853268 Chr.1: 192576412-192576412 on Build GRCh38
rs1039848628 Chr.1: 192576452-192576452 on Build GRCh38
rs542985822 Chr.1: 192576428-192576428 on Build GRCh38
rs367791605 Chr.1: 192576425-192576425 on Build GRCh38
rs995183259 Chr.1: 192576479-192576479 on Build GRCh38
rs770835460 Chr.1: 192576410-192576410 on Build GRCh38
rs371838216 Chr.1: 192576336-192576336 on Build GRCh38
rs896499658 Chr.1: 192576496-192576496 on Build GRCh38
rs778017564 Chr.1: 192576364-192576364 on Build GRCh38
rs375194538 Chr.1: 192576374-192576374 on Build GRCh38
rs1035263429 Chr.1: 192576485-192576485 on Build GRCh38
rs1038978700 Chr.1: 192576468-192576468 on Build GRCh38
rs371716956 Chr.1: 192576378-192576378 on Build GRCh38
rs115518462 Chr.1: 192576480-192576480 on Build GRCh38
rs573246885 Chr.1: 192576435-192576435 on Build GRCh38
rs749496425 Chr.1: 192576390-192576390 on Build GRCh38
rs1046819546 Chr.1: 192576429-192576429 on Build GRCh38
rs780143481 Chr.1: 192576384-192576384 on Build GRCh38
rs530912821 Chr.1: 192576481-192576481 on Build GRCh38
rs568140380 Chr.1: 192576460-192576460 on Build GRCh38

View Full Product Details