Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155256208-155256467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCAMP3
Entrez Gene ID
10067
Gene Name
secretory carrier membrane protein 3
Gene Aliases
C1orf3
UniGene
Hs.200600
Gene Chromosome Location
Chr.1: 155255979-155262703 on Build GRCh38
Transcripts
NM_005698.3 NM_052837.2 XM_006711105.3 XM_006711106.3 XM_016999991.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7176937 c.1032C>T p.F344F Chr.1: 155256285-155256285 on Build GRCh38
6772408 c.913C>T p.R305C Chr.1: 155256404-155256404 on Build GRCh38
5038918 c.1036G>T p.A346S Chr.1: 155256281-155256281 on Build GRCh38
7232286 c.1033C>A p.R345R Chr.1: 155256284-155256284 on Build GRCh38
7239694 c.967T>C p.F323L Chr.1: 155256350-155256350 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765709365 Chr.1: 155256329-155256329 on Build GRCh38
rs749279293 Chr.1: 155256373-155256373 on Build GRCh38
rs373022803 Chr.1: 155256250-155256250 on Build GRCh38
rs768312018 Chr.1: 155256284-155256284 on Build GRCh38
rs115599747 Chr.1: 155256442-155256442 on Build GRCh38
rs954727241 Chr.1: 155256246-155256246 on Build GRCh38
rs757580799 Chr.1: 155256328-155256328 on Build GRCh38
rs34315387 Chr.1: 155256424-155256424 on Build GRCh38
rs147380030 Chr.1: 155256422-155256422 on Build GRCh38
rs888126153 Chr.1: 155256382-155256382 on Build GRCh38
rs746718405 Chr.1: 155256283-155256283 on Build GRCh38
rs754272676 Chr.1: 155256241-155256241 on Build GRCh38
rs747925342 Chr.1: 155256307-155256307 on Build GRCh38
rs760695138 Chr.1: 155256402-155256402 on Build GRCh38
rs989802609 Chr.1: 155256234-155256234 on Build GRCh38
rs754617388 Chr.1: 155256351-155256351 on Build GRCh38
rs2990233 Chr.1: 155256326-155256326 on Build GRCh38
rs1044491531 Chr.1: 155256257-155256257 on Build GRCh38
rs752647282 Chr.1: 155256268-155256268 on Build GRCh38
rs764324101 Chr.1: 155256404-155256404 on Build GRCh38
rs767022885 Chr.1: 155256423-155256423 on Build GRCh38
rs927358409 Chr.1: 155256265-155256265 on Build GRCh38
rs769722791 Chr.1: 155256308-155256308 on Build GRCh38
rs765580566 Chr.1: 155256415-155256415 on Build GRCh38
rs551574329 Chr.1: 155256359-155256359 on Build GRCh38
rs913567708 Chr.1: 155256360-155256360 on Build GRCh38
rs775673954 Chr.1: 155256323-155256323 on Build GRCh38
rs367772504 Chr.1: 155256395-155256395 on Build GRCh38
rs779849544 Chr.1: 155256281-155256281 on Build GRCh38
rs779761545 Chr.1: 155256342-155256342 on Build GRCh38
rs371502011 Chr.1: 155256414-155256414 on Build GRCh38
rs776722756 Chr.1: 155256411-155256411 on Build GRCh38
rs372529313 Chr.1: 155256431-155256431 on Build GRCh38
rs754460678 Chr.1: 155256430-155256430 on Build GRCh38
rs775510683 Chr.1: 155256400-155256400 on Build GRCh38
rs111710029 Chr.1: 155256247-155256247 on Build GRCh38
rs760929707 Chr.1: 155256324-155256324 on Build GRCh38
rs777590989 Chr.1: 155256444-155256444 on Build GRCh38
rs11557755 Chr.1: 155256236-155256236 on Build GRCh38
rs201406357 Chr.1: 155256447-155256447 on Build GRCh38
rs746626454 Chr.1: 155256343-155256343 on Build GRCh38
rs779315685 Chr.1: 155256269-155256269 on Build GRCh38
rs554598948 Chr.1: 155256309-155256309 on Build GRCh38
rs200941865 Chr.1: 155256316-155256316 on Build GRCh38
rs762283187 Chr.1: 155256323-155256324 on Build GRCh38
rs771714139 Chr.1: 155256279-155256279 on Build GRCh38
rs565413473 Chr.1: 155256277-155256277 on Build GRCh38
rs778252831 Chr.1: 155256240-155256240 on Build GRCh38
rs759541815 Chr.1: 155256312-155256312 on Build GRCh38
rs747818902 Chr.1: 155256432-155256432 on Build GRCh38
rs780729430 Chr.1: 155256356-155256356 on Build GRCh38
rs776528808 Chr.1: 155256305-155256305 on Build GRCh38
rs750946574 Chr.1: 155256419-155256419 on Build GRCh38
rs769477147 Chr.1: 155256372-155256372 on Build GRCh38
rs757970705 Chr.1: 155256337-155256337 on Build GRCh38
rs751039504 Chr.1: 155256331-155256331 on Build GRCh38
rs765830038 Chr.1: 155256437-155256438 on Build GRCh38
rs930280834 Chr.1: 155256332-155256332 on Build GRCh38

View Full Product Details