Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 167000260-167000533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPOCK3
Entrez Gene ID
50859
Gene Name
sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 3
Gene Aliases
HSAJ1454, TES-3, TICN3
UniGene
Hs.481133
Gene Chromosome Location
Chr.4: 166733384-167234869 on Build GRCh38
Transcripts
NM_001040159.1 NM_001204352.1 NM_001204353.1 NM_001204354.1 NM_001204355.1 NM_001204356.1 NM_001204358.1 NM_001251967.1 NM_016950.2 XM_011532018.1 XM_017008257.1 XM_017008258.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5442716 c.325A>T p.I109F Chr.4: 167000374-167000374 on Build GRCh38
4330734 c.172G>A p.A58T Chr.4: 167000383-167000383 on Build GRCh38
4005701 c.250G>A p.D84N Chr.4: 167000458-167000458 on Build GRCh38
5995603 c.191G>C p.R64P Chr.4: 167000364-167000364 on Build GRCh38
3335711 c.270G>A p.M90I Chr.4: 167000429-167000429 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1039393138 Chr.4: 167000469-167000469 on Build GRCh38
rs193157802 Chr.4: 167000453-167000453 on Build GRCh38
rs777139850 Chr.4: 167000366-167000366 on Build GRCh38
rs768002472 Chr.4: 167000470-167000470 on Build GRCh38
rs765313716 Chr.4: 167000337-167000337 on Build GRCh38
rs11557045 Chr.4: 167000418-167000418 on Build GRCh38
rs759137530 Chr.4: 167000446-167000446 on Build GRCh38
rs771331099 Chr.4: 167000466-167000466 on Build GRCh38
rs770681322 Chr.4: 167000309-167000309 on Build GRCh38
rs753175621 Chr.4: 167000471-167000471 on Build GRCh38
rs957891655 Chr.4: 167000286-167000286 on Build GRCh38
rs747520442 Chr.4: 167000311-167000311 on Build GRCh38
rs9685645 Chr.4: 167000374-167000374 on Build GRCh38
rs751840995 Chr.4: 167000347-167000347 on Build GRCh38
rs370725678 Chr.4: 167000340-167000340 on Build GRCh38
rs35168179 Chr.4: 167000408-167000408 on Build GRCh38
rs779542712 Chr.4: 167000501-167000501 on Build GRCh38
rs1049800931 Chr.4: 167000480-167000480 on Build GRCh38
rs756678915 Chr.4: 167000327-167000327 on Build GRCh38
rs761831184 Chr.4: 167000334-167000334 on Build GRCh38
rs778379282 Chr.4: 167000494-167000494 on Build GRCh38
rs576645951 Chr.4: 167000486-167000486 on Build GRCh38
rs776992870 Chr.4: 167000401-167000401 on Build GRCh38
rs760610683 Chr.4: 167000415-167000415 on Build GRCh38
rs367853804 Chr.4: 167000310-167000310 on Build GRCh38
rs755427328 Chr.4: 167000359-167000359 on Build GRCh38
rs767408854 Chr.4: 167000321-167000321 on Build GRCh38
rs772945866 Chr.4: 167000300-167000300 on Build GRCh38
rs147127158 Chr.4: 167000365-167000365 on Build GRCh38
rs572553472 Chr.4: 167000443-167000443 on Build GRCh38
rs888022978 Chr.4: 167000505-167000505 on Build GRCh38
rs554177496 Chr.4: 167000409-167000409 on Build GRCh38
rs371965762 Chr.4: 167000478-167000478 on Build GRCh38
rs762512895 Chr.4: 167000307-167000307 on Build GRCh38
rs778390054 Chr.4: 167000476-167000476 on Build GRCh38
rs979793494 Chr.4: 167000331-167000331 on Build GRCh38
rs756671647 Chr.4: 167000472-167000472 on Build GRCh38
rs774090845 Chr.4: 167000397-167000397 on Build GRCh38
rs773144661 Chr.4: 167000419-167000419 on Build GRCh38
rs770464530 Chr.4: 167000396-167000396 on Build GRCh38
rs200466103 Chr.4: 167000457-167000457 on Build GRCh38
rs751832396 Chr.4: 167000452-167000452 on Build GRCh38
rs748682348 Chr.4: 167000364-167000364 on Build GRCh38
rs745532216 Chr.4: 167000504-167000504 on Build GRCh38
rs941239837 Chr.4: 167000410-167000410 on Build GRCh38
rs374065369 Chr.4: 167000341-167000341 on Build GRCh38

View Full Product Details