Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 62074994-62075239 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYT16
Entrez Gene ID
83851
Gene Name
synaptotagmin 16
Gene Aliases
CHR14SYT, SYT14L, Strep14, syt14r, yt14r
UniGene
Hs.404139
Gene Chromosome Location
Chr.14: 61812156-62112825 on Build GRCh38
Transcripts
NM_031914.2 XM_006720272.3 XM_011537219.1 XM_011537220.1 XM_011537221.2 XM_011537222.2 XM_011537223.2 XM_011537224.2 XM_011537225.2 XM_011537227.2 XM_011537228.2 XM_011537229.2 XM_017021693.1 XM_017021694.1 XM_017021695.1 XM_017021696.1 XM_017021697.1 XM_017021698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1152760 c.783C>T p.Y261Y Chr.14: 62075181-62075181 on Build GRCh38
7106315 c.788A>G p.E263G Chr.14: 62075186-62075186 on Build GRCh38
7223717 c.821C>T p.P274L Chr.14: 62075219-62075219 on Build GRCh38
2247301 c.779C>T p.S260F Chr.14: 62075177-62075177 on Build GRCh38
6300314 c.745G>A p.G249R Chr.14: 62075143-62075143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933930965 Chr.14: 62075046-62075046 on Build GRCh38
rs762910262 Chr.14: 62075179-62075179 on Build GRCh38
rs201294056 Chr.14: 62075159-62075159 on Build GRCh38
rs773198773 Chr.14: 62075113-62075113 on Build GRCh38
rs533381915 Chr.14: 62075116-62075116 on Build GRCh38
rs376399420 Chr.14: 62075195-62075195 on Build GRCh38
rs922248753 Chr.14: 62075043-62075043 on Build GRCh38
rs896446193 Chr.14: 62075207-62075207 on Build GRCh38
rs972353488 Chr.14: 62075027-62075027 on Build GRCh38
rs780310096 Chr.14: 62075151-62075151 on Build GRCh38
rs758800471 Chr.14: 62075150-62075150 on Build GRCh38
rs773110798 Chr.14: 62075173-62075173 on Build GRCh38
rs370529985 Chr.14: 62075200-62075200 on Build GRCh38
rs761749682 Chr.14: 62075087-62075087 on Build GRCh38
rs369414744 Chr.14: 62075182-62075182 on Build GRCh38
rs552843156 Chr.14: 62075126-62075126 on Build GRCh38
rs568243731 Chr.14: 62075204-62075204 on Build GRCh38
rs764478920 Chr.14: 62075060-62075060 on Build GRCh38
rs532734247 Chr.14: 62075123-62075123 on Build GRCh38
rs143916565 Chr.14: 62075114-62075114 on Build GRCh38
rs759162330 Chr.14: 62075183-62075183 on Build GRCh38
rs187241107 Chr.14: 62075080-62075080 on Build GRCh38
rs911096147 Chr.14: 62075065-62075065 on Build GRCh38
rs751019852 Chr.14: 62075133-62075133 on Build GRCh38
rs759133953 Chr.14: 62075118-62075118 on Build GRCh38
rs373403641 Chr.14: 62075188-62075188 on Build GRCh38
rs201271787 Chr.14: 62075158-62075158 on Build GRCh38
rs776593873 Chr.14: 62075086-62075086 on Build GRCh38
rs1003753137 Chr.14: 62075093-62075093 on Build GRCh38
rs1050068912 Chr.14: 62075051-62075051 on Build GRCh38
rs372127808 Chr.14: 62075155-62075155 on Build GRCh38
rs752156211 Chr.14: 62075119-62075119 on Build GRCh38
rs754285330 Chr.14: 62075147-62075147 on Build GRCh38
rs377092097 Chr.14: 62075181-62075181 on Build GRCh38
rs915247607 Chr.14: 62075177-62075177 on Build GRCh38
rs769726067 Chr.14: 62075106-62075106 on Build GRCh38
rs766210308 Chr.14: 62075115-62075115 on Build GRCh38
rs763727151 Chr.14: 62075129-62075130 on Build GRCh38
rs760243988 Chr.14: 62075198-62075198 on Build GRCh38
rs758991608 Chr.14: 62075205-62075205 on Build GRCh38
rs766706431 Chr.14: 62075210-62075210 on Build GRCh38
rs768771757 Chr.14: 62075156-62075156 on Build GRCh38
rs763740767 Chr.14: 62075199-62075199 on Build GRCh38
rs548261719 Chr.14: 62075164-62075164 on Build GRCh38
rs1025785386 Chr.14: 62075045-62075053 on Build GRCh38

View Full Product Details