Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 1934678-1934913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRPA
Entrez Gene ID
140885
Gene Name
signal regulatory protein alpha
Gene Aliases
BIT, CD172A, MFR, MYD-1, P84, PTPNS1, SHPS1, SIRP
UniGene
Hs.581021
Gene Chromosome Location
Chr.20: 1894117-1939895 on Build GRCh38
Transcripts
NM_001040022.1 NM_001040023.1 NM_080792.2 XM_005260669.3 XM_005260670.3 XM_006723545.3 XM_011529173.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159691 c.1231C>A p.H411N Chr.20: 1934719-1934719 on Build GRCh38
3404961 c.1241A>T p.E414V Chr.20: 1934729-1934729 on Build GRCh38
4818364 c.1264C>T p.Q422* Chr.20: 1934752-1934752 on Build GRCh38
7389966 c.1253G>T p.R418I Chr.20: 1934741-1934741 on Build GRCh38
7389967 c.1253G>T p.R418I Chr.20: 1934741-1934741 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766738587 Chr.20: 1934819-1934819 on Build GRCh38
rs773875185 Chr.20: 1934786-1934786 on Build GRCh38
rs910963836 Chr.20: 1934797-1934797 on Build GRCh38
rs942398182 Chr.20: 1934811-1934811 on Build GRCh38
rs766644353 Chr.20: 1934802-1934802 on Build GRCh38
rs745645458 Chr.20: 1934744-1934744 on Build GRCh38
rs955216012 Chr.20: 1934747-1934747 on Build GRCh38
rs769637747 Chr.20: 1934746-1934746 on Build GRCh38
rs748895218 Chr.20: 1934753-1934753 on Build GRCh38
rs754073338 Chr.20: 1934804-1934804 on Build GRCh38
rs920315710 Chr.20: 1934870-1934870 on Build GRCh38
rs757172007 Chr.20: 1934727-1934727 on Build GRCh38
rs141454528 Chr.20: 1934728-1934728 on Build GRCh38
rs920192812 Chr.20: 1934824-1934824 on Build GRCh38
rs779634840 Chr.20: 1934718-1934723 on Build GRCh38
rs754682128 Chr.20: 1934840-1934840 on Build GRCh38
rs11906187 Chr.20: 1934858-1934858 on Build GRCh38
rs779843288 Chr.20: 1934750-1934750 on Build GRCh38
rs973668768 Chr.20: 1934818-1934818 on Build GRCh38
rs748580205 Chr.20: 1934853-1934853 on Build GRCh38
rs768288574 Chr.20: 1934754-1934754 on Build GRCh38
rs200228072 Chr.20: 1934718-1934718 on Build GRCh38
rs773122242 Chr.20: 1934792-1934792 on Build GRCh38
rs941650234 Chr.20: 1934817-1934817 on Build GRCh38
rs781685472 Chr.20: 1934841-1934841 on Build GRCh38
rs930206500 Chr.20: 1934850-1934850 on Build GRCh38
rs751114776 Chr.20: 1934839-1934839 on Build GRCh38
rs760829085 Chr.20: 1934793-1934793 on Build GRCh38
rs868458773 Chr.20: 1934759-1934759 on Build GRCh38
rs541111031 Chr.20: 1934707-1934707 on Build GRCh38
rs373369770 Chr.20: 1934798-1934798 on Build GRCh38

View Full Product Details