Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89837530-89837803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3S2
Entrez Gene ID
10239
Gene Name
adaptor related protein complex 3 sigma 2 subunit
Gene Aliases
AP3S3, sigma3b
UniGene
Hs.632161
Gene Chromosome Location
Chr.15: 89830599-89894385 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1

Gene Symbol
C15orf38-AP3S2
Entrez Gene ID
100526783
Gene Name
C15orf38-AP3S2 readthrough
Gene Aliases
-
UniGene
Hs.744890
Gene Chromosome Location
Chr.15: 89830599-89912990 on Build GRCh38
Transcripts
NM_005829.4 NR_023361.1 NR_037582.1 NM_001199058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1678677 c.406G>T p.E136* Chr.15: 89837662-89837662 on Build GRCh38
7294942 c.382A>G p.M128V Chr.15: 89837686-89837686 on Build GRCh38
6036748 c.411C>T p.I137I Chr.15: 89837657-89837657 on Build GRCh38
4611607 c.377delG p.G126fs*9 Chr.15: 89837691-89837691 on Build GRCh38
1183144 c.371T>C p.V124A Chr.15: 89837697-89837697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537215013 Chr.15: 89837643-89837643 on Build GRCh38
rs773548963 Chr.15: 89837681-89837681 on Build GRCh38
rs1020053494 Chr.15: 89837783-89837783 on Build GRCh38
rs899096822 Chr.15: 89837705-89837705 on Build GRCh38
rs760799457 Chr.15: 89837602-89837602 on Build GRCh38
rs767478842 Chr.15: 89837722-89837722 on Build GRCh38
rs368634689 Chr.15: 89837764-89837764 on Build GRCh38
rs753505908 Chr.15: 89837760-89837760 on Build GRCh38
rs753260830 Chr.15: 89837762-89837762 on Build GRCh38
rs771748143 Chr.15: 89837608-89837609 on Build GRCh38
rs775562043 Chr.15: 89837759-89837759 on Build GRCh38
rs370559447 Chr.15: 89837617-89837617 on Build GRCh38
rs750130531 Chr.15: 89837729-89837729 on Build GRCh38
rs758842886 Chr.15: 89837763-89837763 on Build GRCh38
rs757167564 Chr.15: 89837627-89837627 on Build GRCh38
rs776807195 Chr.15: 89837696-89837696 on Build GRCh38
rs555192998 Chr.15: 89837689-89837689 on Build GRCh38
rs368032985 Chr.15: 89837676-89837676 on Build GRCh38
rs772403206 Chr.15: 89837582-89837582 on Build GRCh38
rs984104930 Chr.15: 89837649-89837649 on Build GRCh38
rs747266817 Chr.15: 89837682-89837682 on Build GRCh38
rs573708975 Chr.15: 89837782-89837782 on Build GRCh38
rs766536591 Chr.15: 89837607-89837607 on Build GRCh38
rs569926519 Chr.15: 89837560-89837560 on Build GRCh38
rs774471001 Chr.15: 89837701-89837701 on Build GRCh38
rs375132697 Chr.15: 89837646-89837646 on Build GRCh38
rs748618297 Chr.15: 89837668-89837668 on Build GRCh38
rs920510361 Chr.15: 89837640-89837640 on Build GRCh38
rs926210519 Chr.15: 89837595-89837595 on Build GRCh38
rs769976020 Chr.15: 89837699-89837699 on Build GRCh38
rs761838736 Chr.15: 89837707-89837707 on Build GRCh38
rs375974136 Chr.15: 89837761-89837761 on Build GRCh38
rs764086530 Chr.15: 89837618-89837618 on Build GRCh38
rs879017766 Chr.15: 89837717-89837717 on Build GRCh38
rs917326756 Chr.15: 89837648-89837648 on Build GRCh38
rs536449634 Chr.15: 89837712-89837712 on Build GRCh38
rs759488352 Chr.15: 89837698-89837698 on Build GRCh38
rs748462682 Chr.15: 89837580-89837580 on Build GRCh38
rs776514378 Chr.15: 89837608-89837608 on Build GRCh38
rs750158643 Chr.15: 89837638-89837638 on Build GRCh38
rs1047049744 Chr.15: 89837659-89837659 on Build GRCh38
rs145323466 Chr.15: 89837641-89837641 on Build GRCh38
rs747298106 Chr.15: 89837772-89837772 on Build GRCh38
rs774727369 Chr.15: 89837565-89837565 on Build GRCh38
rs758691462 Chr.15: 89837687-89837687 on Build GRCh38
rs760935591 Chr.15: 89837657-89837657 on Build GRCh38
rs146312709 Chr.15: 89837620-89837620 on Build GRCh38
rs773458887 Chr.15: 89837587-89837587 on Build GRCh38
rs1029271600 Chr.15: 89837741-89837741 on Build GRCh38
rs1049968628 Chr.15: 89837781-89837781 on Build GRCh38
rs371466465 Chr.15: 89837747-89837747 on Build GRCh38
rs767390636 Chr.15: 89837634-89837634 on Build GRCh38

View Full Product Details