Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 31224137-31224386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIM72
Entrez Gene ID
493829
Gene Name
tripartite motif containing 72
Gene Aliases
MG53
UniGene
Hs.661859
Gene Chromosome Location
Chr.16: 31214021-31225656 on Build GRCh38
Transcripts
NM_001008274.3 XM_017023254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4774482 c.913T>C p.S305P Chr.16: 31224234-31224234 on Build GRCh38
1290514 c.974C>A p.P325Q Chr.16: 31224295-31224295 on Build GRCh38
7222766 c.871C>T p.L291L Chr.16: 31224192-31224192 on Build GRCh38
3420958 c.975G>A p.P325P Chr.16: 31224296-31224296 on Build GRCh38
6808600 c.1023C>T p.S341S Chr.16: 31224344-31224344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756750610 Chr.16: 31224344-31224344 on Build GRCh38
rs200555243 Chr.16: 31224194-31224194 on Build GRCh38
rs751349807 Chr.16: 31224212-31224212 on Build GRCh38
rs755691030 Chr.16: 31224220-31224220 on Build GRCh38
rs952160847 Chr.16: 31224353-31224353 on Build GRCh38
rs866681890 Chr.16: 31224243-31224243 on Build GRCh38
rs773218230 Chr.16: 31224280-31224280 on Build GRCh38
rs900182053 Chr.16: 31224162-31224162 on Build GRCh38
rs112486041 Chr.16: 31224288-31224288 on Build GRCh38
rs989532506 Chr.16: 31224297-31224297 on Build GRCh38
rs764242730 Chr.16: 31224348-31224350 on Build GRCh38
rs767357835 Chr.16: 31224317-31224317 on Build GRCh38
rs368859412 Chr.16: 31224247-31224247 on Build GRCh38
rs58585879 Chr.16: 31224227-31224227 on Build GRCh38
rs369457250 Chr.16: 31224332-31224332 on Build GRCh38
rs770809630 Chr.16: 31224308-31224308 on Build GRCh38
rs773204133 Chr.16: 31224193-31224193 on Build GRCh38
rs773058167 Chr.16: 31224264-31224264 on Build GRCh38
rs754784371 Chr.16: 31224208-31224208 on Build GRCh38
rs201970764 Chr.16: 31224333-31224333 on Build GRCh38
rs752343664 Chr.16: 31224219-31224219 on Build GRCh38
rs774300796 Chr.16: 31224312-31224312 on Build GRCh38
rs1003218502 Chr.16: 31224283-31224283 on Build GRCh38
rs761902597 Chr.16: 31224164-31224164 on Build GRCh38
rs372493107 Chr.16: 31224284-31224284 on Build GRCh38
rs781404405 Chr.16: 31224263-31224263 on Build GRCh38
rs375401025 Chr.16: 31224313-31224313 on Build GRCh38
rs1019552922 Chr.16: 31224249-31224249 on Build GRCh38
rs767302913 Chr.16: 31224211-31224211 on Build GRCh38
rs753329636 Chr.16: 31224338-31224338 on Build GRCh38
rs995214440 Chr.16: 31224167-31224167 on Build GRCh38
rs199815688 Chr.16: 31224324-31224324 on Build GRCh38
rs372288148 Chr.16: 31224251-31224251 on Build GRCh38
rs936743874 Chr.16: 31224293-31224293 on Build GRCh38
rs754762140 Chr.16: 31224227-31224227 on Build GRCh38
rs766397238 Chr.16: 31224202-31224216 on Build GRCh38
rs765240001 Chr.16: 31224165-31224165 on Build GRCh38
rs1046980046 Chr.16: 31224364-31224364 on Build GRCh38
rs534421116 Chr.16: 31224204-31224204 on Build GRCh38
rs150087468 Chr.16: 31224196-31224196 on Build GRCh38
rs141229511 Chr.16: 31224258-31224258 on Build GRCh38
rs763780493 Chr.16: 31224337-31224337 on Build GRCh38
rs375350426 Chr.16: 31224275-31224275 on Build GRCh38
rs571127764 Chr.16: 31224277-31224277 on Build GRCh38
rs758879061 Chr.16: 31224246-31224246 on Build GRCh38

View Full Product Details