Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 159951473-159951742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLAMF9
Entrez Gene ID
89886
Gene Name
SLAM family member 9
Gene Aliases
CD2F-10, CD2F10, CD84-H1, CD84H1, SF2001
UniGene
Hs.661712
Gene Chromosome Location
Chr.1: 159951492-159984730 on Build GRCh38
Transcripts
NM_001146172.1 NM_001146173.1 NM_033438.3 XM_017002756.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2084981 c.809A>C p.K270T Chr.1: 159951722-159951722 on Build GRCh38
5353014 c.797C>A p.P266Q Chr.1: 159951734-159951734 on Build GRCh38
4951695 c.823A>G p.R275G Chr.1: 159951708-159951708 on Build GRCh38
1688931 c.828G>A p.M276I Chr.1: 159951703-159951703 on Build GRCh38
5700098 c.837G>T p.R279S Chr.1: 159951694-159951694 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754461680 Chr.1: 159951640-159951640 on Build GRCh38
rs747570690 Chr.1: 159951670-159951670 on Build GRCh38
rs1026258566 Chr.1: 159951513-159951513 on Build GRCh38
rs189527068 Chr.1: 159951655-159951655 on Build GRCh38
rs141914707 Chr.1: 159951630-159951630 on Build GRCh38
rs370490018 Chr.1: 159951669-159951669 on Build GRCh38
rs765413043 Chr.1: 159951615-159951615 on Build GRCh38
rs747643397 Chr.1: 159951646-159951646 on Build GRCh38
rs1023999665 Chr.1: 159951571-159951571 on Build GRCh38
rs926605607 Chr.1: 159951553-159951553 on Build GRCh38
rs753988317 Chr.1: 159951610-159951610 on Build GRCh38
rs748732294 Chr.1: 159951561-159951561 on Build GRCh38
rs755738878 Chr.1: 159951647-159951647 on Build GRCh38
rs755631790 Chr.1: 159951548-159951548 on Build GRCh38
rs779902020 Chr.1: 159951625-159951625 on Build GRCh38
rs938113472 Chr.1: 159951554-159951554 on Build GRCh38
rs370276711 Chr.1: 159951644-159951644 on Build GRCh38
rs751465647 Chr.1: 159951706-159951706 on Build GRCh38
rs765442763 Chr.1: 159951683-159951683 on Build GRCh38
rs377218732 Chr.1: 159951663-159951663 on Build GRCh38
rs766208517 Chr.1: 159951697-159951697 on Build GRCh38
rs142608001 Chr.1: 159951677-159951677 on Build GRCh38
rs148110395 Chr.1: 159951529-159951529 on Build GRCh38
rs200096062 Chr.1: 159951692-159951692 on Build GRCh38
rs746173192 Chr.1: 159951649-159951649 on Build GRCh38
rs569805928 Chr.1: 159951611-159951611 on Build GRCh38
rs1000322597 Chr.1: 159951494-159951494 on Build GRCh38
rs202043270 Chr.1: 159951688-159951688 on Build GRCh38
rs915299946 Chr.1: 159951583-159951583 on Build GRCh38
rs768829492 Chr.1: 159951671-159951671 on Build GRCh38
rs758853588 Chr.1: 159951677-159951677 on Build GRCh38
rs757043346 Chr.1: 159951574-159951574 on Build GRCh38
rs758203661 Chr.1: 159951620-159951620 on Build GRCh38
rs970629832 Chr.1: 159951522-159951522 on Build GRCh38
rs1007733749 Chr.1: 159951699-159951699 on Build GRCh38
rs772440315 Chr.1: 159951597-159951597 on Build GRCh38
rs762734108 Chr.1: 159951694-159951694 on Build GRCh38
rs1014420172 Chr.1: 159951564-159951564 on Build GRCh38
rs777301322 Chr.1: 159951648-159951648 on Build GRCh38
rs184005867 Chr.1: 159951509-159951509 on Build GRCh38
rs750183545 Chr.1: 159951618-159951618 on Build GRCh38

View Full Product Details