Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149776880-149777107 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SSPO
Entrez Gene ID
23145
Gene Name
SCO-spondin
Gene Aliases
-
UniGene
Hs.632022
Gene Chromosome Location
Chr.7: 149776042-149833965 on Build GRCh38
Transcripts
NM_198455.2

SNP Detail

SNP ID Chromosome Location
rs776510770 Chr.7: 149777043-149777043 on Build GRCh38
rs772131287 Chr.7: 149776994-149776994 on Build GRCh38
rs373910612 Chr.7: 149776947-149776947 on Build GRCh38
rs770908881 Chr.7: 149777042-149777042 on Build GRCh38
rs757443911 Chr.7: 149776933-149776933 on Build GRCh38
rs780380699 Chr.7: 149777003-149777003 on Build GRCh38
rs73727604 Chr.7: 149776910-149776910 on Build GRCh38
rs371869171 Chr.7: 149777046-149777046 on Build GRCh38
rs796848519 Chr.7: 149776988-149776988 on Build GRCh38
rs368470785 Chr.7: 149777002-149777002 on Build GRCh38
rs559860257 Chr.7: 149777036-149777036 on Build GRCh38
rs767489947 Chr.7: 149777054-149777054 on Build GRCh38
rs760599757 Chr.7: 149776991-149776991 on Build GRCh38
rs137982924 Chr.7: 149777048-149777048 on Build GRCh38
rs748030684 Chr.7: 149776993-149776994 on Build GRCh38
rs374321064 Chr.7: 149777033-149777033 on Build GRCh38
rs772907780 Chr.7: 149776987-149776987 on Build GRCh38
rs201087823 Chr.7: 149776973-149776973 on Build GRCh38
rs376137620 Chr.7: 149776919-149776919 on Build GRCh38
rs544783107 Chr.7: 149776921-149776921 on Build GRCh38
rs186147601 Chr.7: 149777044-149777044 on Build GRCh38
rs749805941 Chr.7: 149777021-149777021 on Build GRCh38
rs755444057 Chr.7: 149777028-149777028 on Build GRCh38
rs564966313 Chr.7: 149776966-149776966 on Build GRCh38
rs367648284 Chr.7: 149777045-149777045 on Build GRCh38
rs575484984 Chr.7: 149776968-149776968 on Build GRCh38
rs767225964 Chr.7: 149776904-149776904 on Build GRCh38
rs751789636 Chr.7: 149776932-149776932 on Build GRCh38
rs372204042 Chr.7: 149777030-149777030 on Build GRCh38
rs1020803656 Chr.7: 149776917-149776917 on Build GRCh38
rs773850970 Chr.7: 149776967-149776967 on Build GRCh38
rs759534249 Chr.7: 149776996-149776996 on Build GRCh38
rs200197861 Chr.7: 149776935-149776935 on Build GRCh38
rs1047076643 Chr.7: 149776936-149776936 on Build GRCh38
rs1051836617 Chr.7: 149777080-149777080 on Build GRCh38
rs368078874 Chr.7: 149776949-149776949 on Build GRCh38
rs780577067 Chr.7: 149776948-149776948 on Build GRCh38
rs890947078 Chr.7: 149776908-149776908 on Build GRCh38
rs779005942 Chr.7: 149776901-149776903 on Build GRCh38
rs180803695 Chr.7: 149776920-149776920 on Build GRCh38
rs529229055 Chr.7: 149776994-149776994 on Build GRCh38
rs761510708 Chr.7: 149776901-149776901 on Build GRCh38
rs550946943 Chr.7: 149777077-149777077 on Build GRCh38
rs561237413 Chr.7: 149776980-149776980 on Build GRCh38
rs549410517 Chr.7: 149777034-149777034 on Build GRCh38
rs998585851 Chr.7: 149777005-149777005 on Build GRCh38
rs764232026 Chr.7: 149776923-149776923 on Build GRCh38
rs767828076 Chr.7: 149777000-149777000 on Build GRCh38
rs772989628 Chr.7: 149776909-149776909 on Build GRCh38
rs374297997 Chr.7: 149776999-149776999 on Build GRCh38

View Full Product Details